+
For Patients & Caregivers
For Healthcare Professionals
Welcome! Click here for Patient or Healthcare Professional content
Are you a healthcare professional?
The information in this website is intended only for healthcare professionals. By entering this site, you are confirming that you are a healthcare professional.
Are you a laboratory professional?
The information in this website is intended only for laboratory professionals. By entering this site, you are confirming that you are a laboratory professional.
EliA Assays
Designed to offer your laboratory high performance and reliable results
Based on advanced techniques such as recombinant antigen technology, EliA™ assays are standardized, precise, reliable and easy-to-use. We currently offer autoantibody tests for more than 20 clinical indications in the fully automated EliA system.
One unique aspect of EliA is the ability to utilize one master isotype curve for each group of assays.
All assays that are IgG in nature run from one IgG master curve. The same is true for IgM and IgA–each using a single IgM or IgA master calibration curve. This technology optimizes workflow by eliminating the need to run calibration curves for individual assays. In this case, less is truly more. The benefit of this technology for your laboratory is the ability to optimize workflow by eliminating the need to run calibration curves for every single analyte.
Why specificity matters in autoimmune disease diagnosis
When your laboratory provides highly specific tests, such as the EliA assays, the results matter to clinicians because of their higher positive predictive value, leading to a greater confidence in diagnosis.
EliA assays pioneered the use of recombinant sources, respectively expressed in baculovirus insect cells, in order to secure reproducible and high-quality antigens. This recombinant expression allows the production of large, reproducible batches of purified proteins that overcome the quantity limitations associated with human raw material. As a result, EliA assays are associated with less contamination and fewer false positives.
Native TPO
recombinant TPO
EliA autoimmunity assays
Combining exceptional specificity and sensitivity to aid in autoimmune disease diagnosis
EliA assays, which run on most Phadia™ Laboratory Systems, can help your laboratory aid in the diagnosis of autoimmune diseases including:
  • Antiphospholipid Syndrome
  • Connective Tissue Diseases
  • Gastrointestinal Diseases
  • Celiac Disease
  • Liver Disease
  • Rheumatoid Arthritis
  • Thyroid Diseases
  • Vasculitis
And if your laboratory is already running ImmunoCAP™ Specific IgE blood tests, adding EliA assays is easy.
autoimmune
Prevalence, testing and treatment overviews
ImmunoCAP Elia
Run ImmunoCAP and EliA assays on a single platform
stay up to date
Keep up-to-date on the latest news and research
See the complete list