+
For Patients & Caregivers
For Healthcare Professionals
Welcome! Click here for Patient or Healthcare Professional content
Are you a healthcare professional?
The information in this website is intended only for healthcare professionals. By entering this site, you are confirming that you are a healthcare professional.
Are you a laboratory professional?
The information in this website is intended only for laboratory professionals. By entering this site, you are confirming that you are a laboratory professional.
ImmunoCAP Whole Allergens: Assays That Provide High Sensitivity in Detecting IgE to The Allergen Source
Brought to you by ThermoFisher Scientific
ImmunoCAP Whole Allergens
One allergen source per ImmunoCAP test
These fully quantitative assays provide high sensitivity in detecting IgE to the allergen source. The technology that makes this detection possible is the very high capacity of the cellulose sponge solid matrix of binding proteins. As a result, whole allergens can be suitable as support to rule in an IgE-mediated allergy. Single ImmunoCAPTM Whole Allergen tests are generally more sensitive than mixes of whole allergens, providing quantitative results rather than a qualitative result produced by a mix used for screening.
whole allergens
whole Allergens
ImmunoCAP Whole Allergens contain most proteins and allergen components from the source allergen, for a total of more than 600 allergens including:
Cat (e1)
Animal Dander
Food
Foods
Grass pollen
Grass Pollen
Drugs
Drugs
insect venom
Insect Venoms
tree pollen
Tree Pollen
House dust mite
Mites
Mold - A. alternate (m6)
Mold
occupational allergens
Occupational allergens