+
For Patients & Caregivers
For Healthcare Professionals
Welcome! Click here for Patient or Healthcare Professional content
Are you a healthcare professional?
The information in this website is intended only for healthcare professionals. By entering this site, you are confirming that you are a healthcare professional.
Are you a laboratory professional?
The information in this website is intended only for laboratory professionals. By entering this site, you are confirming that you are a laboratory professional.
ImmunoCAP ISAC
When you need the bigger picture in allergy sensitization
The ImmunoCAPTM ISACTM test is a widely used in vitro diagnostic test that allows for simultaneous measurement of specific IgE antibodies to a broad spectrum of allergen components.
ImmunoCAP ISAC is an immunoassay platform that allows for simultaneous measurement of specific IgE antibodies to 112 different allergen components using only 30 μl of serum or plasma. Capillary blood sampling can be used, thus enabling a less invasive procedure for testing young children.
Purified natural or recombinant molecular allergen components are immobilized on a solid support (biochip). ImmunoCAP ISAC is a semi-quantitative test. Results are reported in ISAC Standardized Units (ISU).
This innovative test provides clinicians with a large amount of allergen specific IgE antibody information in one single step. ImmunoCAP ISAC test is the result of a combination of innovative biochip technology and cutting-edge research in molecular allergology.
NOTE: ImmunoCAP ISAC does not run on Phadia™ Laboratory Systems. ImmunoCAP ISAC testing is available in the US through the Phadia Immunology Reference Laboratory (PiRL)
Give Your Clinicians New Insights
At times, your clinicians will face diagnostic challenges, such as symptomatic patients whose symptoms and case history are inconsistent, who are multisensitized or who have an unsatisfactory response to initial treatment. In these cases, ImmunoCAP ISAC tests can help your laboratory add clinical value by revealing:
  • The sensitization profile of multisensitized patients (ie, cross-reactivity vs co-sensitization)
  • Results that can help a clinician identify potential risks for reaction and type of reaction, especially food-related reactions
  • Unexpected sensitizations
  • The IgE antibody profile in patients with unsatisfactory response to treatment
  • Non-atopics
A Technological Step Forward
Based on modern biochip technology, ImmunoCAP ISAC tests are an immunoassay platform in which allergen components are immobilized in a microarray. This test is a two-step assay that can be completed in less than 4 hours:
  1. IgE antibodies from the patient sample bind to the immobilized allergen components
  2. Allergen-bound IgE antibodies are detected by a fluorescence labeled anti-IgE antibody
ImmunoCAP ISAC tests are semiquantitative, with results reported in ISAC Standardized Units for specific IgE (ISU-E). These data give indications of specific IgE antibody levels within a measuring range of 0.3 to 100 ISU-E.
Key protein families assayed include:
  • Storage protein
  • LTP (nonspecific Lipid Transfer Protein, nsLTP)
  • PR10 protein, Bet v 1 homologue
  • Profilin
  • Cross-reactive carbohydrate determinants (CCDs)
  • Tropomyosin
  • Parvalbumin
  • Serum albumin
Contact
Is Your Laboratory Ready to Make Advanced Technology Part of Your Offerings?
If your laboratory has questions about adding ImmunoCAP ISAC, we can help you.