+
For Patients & Caregivers
For Healthcare Professionals
Welcome! Click here for Patient or Healthcare Professional content
Are you a healthcare professional?
The information in this website is intended only for healthcare professionals. By entering this site, you are confirming that you are a healthcare professional.
Are you a laboratory professional?
The information in this website is intended only for laboratory professionals. By entering this site, you are confirming that you are a laboratory professional.
Phadia 5000 Instrument
Sophisticated and powerful to meet the demands of any major laboratory
Phadia™ 5000 instrument provides the highest throughput amongst the Phadia Laboratory Systems family. With four parallel process lines, it doubles the capacity of Phadia 2500 instrument and provides even greater flexibility. There are three configurations of Phadia 5000 instrument that reflect your assay needs; ImmunoCAP™ only, EliA™ only, or a combination of ImmunoCAP and EliA. This offers your laboratory the ability to tailor a high-throughput automated solution that aligns with your assay demand and schedule.
Ideal for extremely high throughput laboratories that deliver 25,000–45,000 results per week
The Phadia 5000 instrument delivers a range of benefits, all with the capability and functionality that makes sense for your laboratory:
Flexibility
Capability
  • Capacity per process line: 240 results/hour (ImmunoCAP)
  • Capacity per process line: 150 results/hour (EliA)
  • Continuous random access
  • All reagents stored on board
  • 28-day storage of calibration curves
  • Onboard dilution (EliA)
  • Clustered set-up for increased capacity
Reliability
Phadia 5000 Instrument Features
  1. Rack input/output module (160 racks) — Loading area for patient samples, quality controls, ImmunoCAP and EliA wells, calibrators/curve controls
  2. Pipette tip rack input
  3. Cooled ImmunoCAP and EliA well carrier storage (1800 positions)
  4. Development storage (2 x 2 positions)
  5. Cooled conjugate positions (32 positions) automatic barcode reading
  6. Barcode reader for traceability of reagent lot number
  7. LCD for instrument management software
  8. Wash tank, automatic refill function
  9. Handheld barcode reader for traceability of reagent lot number
  10. Stop solution position (2 positions)
  11. Washing solution concentrate and washing solution additive bottles
Not shown in picture:
  1. Fixed pipe connections for purified water and drain
  2. Stand-alone PC for Phadia Prime software, connectable to mainframe Laboratory Information Systems (LIS)
Phadia 5000 Instrument Capability
Phadia 5000
Direction of Use
Directions for Use
(DFU)
Find general directions on
how to use.
Certificate of Analysis
Certificate of Analysis
(CoA)
Access important reference materials for your lab. 
System Safety Data
Material Safety Data
Sheets (MSDS)
Review available safety data sheets for specific products.
Understanding allergic diseases
Are you ready to offer four process lines and even greater flexibility?
If your laboratory has been considering upgrading or adding to your testing system, Thermo Fisher Scientific can help you find the right Phadia Laboratory System for your needs.