WesternDot® secondary antibody detection workflow
WesternDot® secondary antibodies can be used in existing Western Blot protocols. Fluorescent western detection is more convenient than chemiluminescence (ECL) methods. Blots are ready to image immediately after the secondary antibody application. There are no additional substrates or enzymatic/timing concerns, and the fluorescent signals are stable indefinitely.
WesternDot® secondary antibodies can be used in existing  Western Blot protocols. Fluorescent western detection is more convenient than chemiluminescence (ECL) methods. Blots are ready to image immediately after the secondary antibody application. There are no additional substrates or enzymatic/timing concerns, and the fluorescent signals are stable indefinitely.
Related Products
Related Images