MAS Quality Controls (MAS QC)
New: Thermo Scientific™ MAS™ Omni•IMMUNE Max and MAS Omni•IMMUNE PRO Max
More with MAS controls
Do more with less steps, less waste, less cost. Thermo Scientific MAS Omni controls offer highly consolidated solutions to help you simplify QC routine with fewer bottles. Our long shelf life and open vial stability helps streamline QC management and reduce waste.
MAS QC products deliver accurate results to help you ensure reliable patient results. Find a full range of high-quality QC products for general chemistry, immunoassays, cardiac, toxicology, urinalysis, specialty, and more.
For molecular QC solutions, explore Thermo Scientific AcroMetrix Quality Controls and standards. 
Thermo Scientific LabLink xL Laboratory quality control software
Simplify your quality control management with Lablink xL software. This cloud-based software provides real-time peer comparisons, automatic data entry, audit-friendly reports, and much more.
computer
Explore more capabilities with MAS Quality Controls (QC)
More efficiency
Thermo Scientific MAS Omni Quality Controls consolidate routinely used QC products, covering a large number of analytes into a single vial. These consolidated controls improve efficiency while maintaining reliable results.
More performance
Industry-leading up to 5 years lot shelf life*
  • Fewer lot-to-lot validation
  • Benefit from long-term QC management
  • Increase productivity
More time
Once thawed, MAS quality controls are ready to use, straight from  refrigeration. Spend less time every day preparing and more time focusing on patient samples.
  • No reconstitution
  • No need to bring to room temperature.
  • Reduce potential errors
  • Maximize productivity
Minimize waste
Most MAS QC provide 30-day open-vial stability*
  • Minimize waste and maximize usage with better open vial stability
Discover MAS QC products for clinical laboratories
Choose the MAS quality controls you need, each with long open vial stability, a ready-to-use design, and extended shelf life. 
Explore QC related resources
Enhance your knowledge of quality controls by exploring available resources, including scientific webinars, brochures, technical notes, scientific posters, and more. 
Not all products are CE marked or have FDA clearance for sale in the U.S. Availability of products in each country depends on local regulatory marketing authorization status.