Invitrogen
Analytical sensitivity
20 pg/mL
Assay range
20-5,000 pg/mL
Sample type/volume
Supernatant
50 µL
Plasma
50 µL
Serum
50 µL
Hands-on time
1 hr 20 min
Time-to-result
4 hr 45 min
Homogenous (no wash)
No
Interassay CV
<12%
Intraassay CV
<10%
Instrument
Colorimetric Microplate Reader
Product size
5 x 96 Tests
Contents
Pre-coated 96 well plate
Standard
Assay Diluent concentrate
Biotinylated Detection Antibody
SAV-HRP
Wash Buffer
Chromogen
Stop Solution
Adhesive Plate Covers
Shipping condition
Wet or Dry Ice
Storage
2-8°C
Protein name
TIMP2
Protein aliases
Metalloproteinase inhibitor 2, TIMP-2, tissue inhibitor of metalloproteinase 2, Tissue inhibitor of metalloproteinases 2
Species (tested)
Rat
Assay kit format
Sandwich ELISA Kit
Detector antibody conjugate
Biotin
Label or dye
HRP
Gene aliases
Timp-2, Timp2
Gene ID
Gene symbol
Timp2
UniProt ID
Rat TIMP-2 quantitates rat TIMP-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat TIMP-2.
TIMP-1 and TIMP-2 are inhibitory enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP-2 shows the highest binding affinity to both the latent (pro) and active forms of 72 kDa Type IV collagenase (also known as MMP-2 or gelatinase A). It also has affinity for the active form of 92 kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMP-2 inhibits the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support