Precise, reliable and flexible AST with the latest antimicrobials  
Precise, reliable and flexible AST with the latest antimicrobials  
To effectively combat antimicrobial resistance, you need the right antimicrobials at the right dose. To improve patient care outcomes, you need faster, more reliable antimicrobial susceptibility testing (AST) devices. Whether you prefer manual or automated solutions, you’ll find the right fit for your specific susceptibility testing needs and volume requirements with the Thermo Scientific™ Sensititre™ ID/AST System.
The Sensititre System is a scalable and flexible AST solution, accommodating microbiology laboratories of all sizes, which utilizes both in vitro diagnostic (IVD) and research use only (RUO), 96-well microtitre plates for broth microdilution. The Sensititre System can help the microbiology lab provide results that reduce uncertainty for critical care decision makers, promote antibiotic de-escalation and antibiotic stewardship, and help reduce pharmacy costs.
Make confident decisions with a true MIC for AST
Obtaining a precise minimum inhibitory concentration (MIC) ensures the correct dosage of an antimicrobial or antibiotic to limit resistance. Some AST systems calculate the MIC value by extrapolating growth curves and applying dedicated algorithms. The Sensititre System generates true MIC results based on the actual growth of the organism. A true MIC reveals whether a bacterial isolate is susceptible to a particular drug, how susceptible the organism is and the exact point at which the isolate becomes resistant. When comparing your MIC results against the latest clinical breakpoints from EUCAST and CLSI, you can trust the value of a true MIC to deliver the accuracy required for optimal patient outcomes, and to track emerging resistance.
Consolidate your susceptibility testing on a single platform
With over 300 antimicrobials available in extended dilution ranges, the Sensititre System allows you to consolidate confirmatory tests while meeting FDA, CLSI and EUCAST breakpoint requirements.
Thermo Scientific™ Sensititre™ Nephelometer—a simple solution to standardize inoculum density.
Thermo Scientific™ Sensititre™ 8-Channel Programmable Pipette—quick and accurate manual inoculation of microtitre plates, with enhanced ergonomics.
Thermo Scientific™ Sensititre AIM™ Automated Inoculation Delivery System—quickly and accurately doses Sensititre 96-well plates, eliminating both skipped wells and costly repeat tests.
Thermo Scientific™ Sensititre™ ARIS™ 2X Instrument—includes a heated carousel to individually incubate 64 MIC, breakpoint or identification plates, ensuring optimal growth conditions and eliminating repeat tests.
(Not available in Europe)
Thermo Scientific™ Sensititre™ Manual Viewer—perform simple visual reads of your 96-well microtitre plates with our mirrored viewbox.
Thermo Scientific™ Sensititre™ Vizion™ Digital MIC Viewing System—combines automation with visual results, to consolidate all offline testing on a single instrument, with the benefits of Expert System software and LIS connectivity.
Thermo Scientific™ Sensititre™ OptiRead™ Automated Fluorometric Plate Reading System—utilizes fluorescence technology to provide fast, accurate plate reads, and eliminate manual reading.
Thermo Scientific™ Sensititre™ ARIS™ 2X Instrument—includes a heated carousel to individually incubate 64 MIC, breakpoint or identification plates, ensuring optimal growth conditions and eliminating repeat tests.
(Not available in Europe)
Thermo Scientific™ Sensititre™ SWIN™ Software System, Complete—combines results from manual, semi-automated and fully automated read options on a single software platform, to easily consolidate your entire test program and enhance data entry.
Thermo Scientific™ Sensititre™ SWIN™ Software Epidemiology Module—allows users to generate complete, real-time reports and bar graphs in just minutes, including multiple search options to create comprehensive reports specific to your patient population to fully support antimicrobial stewardship programs.
Thermo Scientific™ Sensititre™ Nephelometer—a simple solution to standardize inoculum density.
Thermo Scientific™ Sensititre™ 8-Channel Programmable Pipette—quick and accurate manual inoculation of microtitre plates, with enhanced ergonomics.
Thermo Scientific™ Sensititre AIM™ Automated Inoculation Delivery System—quickly and accurately doses Sensititre 96-well plates, eliminating both skipped wells and costly repeat tests.
Thermo Scientific™ Sensititre™ ARIS™ 2X Instrument—includes a heated carousel to individually incubate 64 MIC, breakpoint or identification plates, ensuring optimal growth conditions and eliminating repeat tests.
(Not available in Europe)
Thermo Scientific™ Sensititre™ Manual Viewer—perform simple visual reads of your 96-well microtitre plates with our mirrored viewbox.
Thermo Scientific™ Sensititre™ Vizion™ Digital MIC Viewing System—combines automation with visual results, to consolidate all offline testing on a single instrument, with the benefits of Expert System software and LIS connectivity.
Thermo Scientific™ Sensititre™ OptiRead™ Automated Fluorometric Plate Reading System—utilizes fluorescence technology to provide fast, accurate plate reads, and eliminate manual reading.
Thermo Scientific™ Sensititre™ ARIS™ 2X Instrument—includes a heated carousel to individually incubate 64 MIC, breakpoint or identification plates, ensuring optimal growth conditions and eliminating repeat tests.
(Not available in Europe)
Thermo Scientific™ Sensititre™ SWIN™ Software System, Complete—combines results from manual, semi-automated and fully automated read options on a single software platform, to easily consolidate your entire test program and enhance data entry.
Thermo Scientific™ Sensititre™ SWIN™ Software Epidemiology Module—allows users to generate complete, real-time reports and bar graphs in just minutes, including multiple search options to create comprehensive reports specific to your patient population to fully support antimicrobial stewardship programs.
Find the Sensititre plates to meet your AST needs
Human
Gram Negative plates
Gram Positive plates
Yeast plates
Mycobacteria plates
Surveillance
Harmonize surveillance AST with plates for AMR programs
View plates
Veterinary
Over 40 veterinary specific antimicrobials and host animal-specific AST plates
View plates
Sensititre Plate Guide
Sensititre Plate Guide
Download our guide to the full range of Sensititre AST plates and layouts in one document.
Download guide
Resources