Jars of salsa ready for checkweighing
Drive process efficiency without skipping a beat
Weigh, count and reject in a single, non-stop checkweigher system. Weigh large, heavy packages without stopping or recalibrating the conveyor. With a Thermo Scientific™ checkweigher customized to your specifications, you can depend on accurate weight control, maximized efficiency, and consistent product throughput, even in rugged industrial environments. From raw or frozen products, bags, cases or barrels to mailers, totes and cases, our high-rate checkweighers will keep your line moving toward maximum productivity at all times.
Contact an application specialist ›    Join our mailing list ›
Popular checkweigher systems
Other checkweighers and in-line scales
Drive down total cost of ownership for dry package weighing applications with the fixed height Thermo Scientific™ Versa Flex Checkweigher. This general purpose unit weighs light to heavy-weight packaged food, pharmaceutical and personal care products. 
Improve accuracy and decrease maintenance and product giveaway with the Thermo Scientific™ Versa Flex GP Checkweigher. This height-adjustable cantilever unit weighs a wide range of packaged food, pharmaceutical and personal care products in dry environments.
Meet the high performance standards of the pharmaceutical industry with the Thermo Scientific™ Versa RxV Marking and Verification Pharmaceutical Checkweigher for track and trace applications and the Versa Rx Pharmaceutical Checkweigher. 
Improve throughput with the Thermo Scientific™ Versa 8120 Chain Checkweigher. With line speeds up to 700 packages per minute, the Versa 8120 Chain Checkweigher provides reliable high-speed weighing, control, and reject functions for cartons, cans, bottles and pouches in dry or wet environments.
Maximize your productivity with the durable and dependable Thermo Scientific™ Versa Frame 44HB Heavy Duty Weighframe, designed for rugged industrial environments with line rates up to 100 packages per minute (up to 300 fpm depending on package length).
Perform dependable and accurate in-line weighing of cases, bags, and large packages up to 110 lb (50 kg) with the Thermo Scientific™ Large Case and Bag Checkweigher - Versa Frame 44 and Box-Series.
Improve efficiency and profitability with a checkweigher designed for better accuracy and faster speeds. The Thermo Scientific™ Global Versa GP Checkweigher is a versatile, general purpose checkweigher that scans at line speeds up to 350 packages per minute without ever sacrificing accuracy.
Maximize profits and minimize product giveaway with the Thermo Scientific™ Global VersaWeigh™ Checkweigher. This versatile, scalable system is easy-to-use and maintain and now features more than 10 engineering and functionality improvements.
Experience unparalleled speed and accuracy in today's demanding, high-rate canning lines with the Thermo Scientific™ Versa Teorema Checkweigher. Combining rugged capability with accuracy, the Versa Teorema checkweigher provides a new standard in high-rate can weighing.
Eliminate the expense of two machines. The Thermo Scientific™ Combination Checkweigher and Metal Detector gives you both accurate weight control and the assurance that your product is free of metal contaminants—all in a single space-saving instrument.
checkweighing and checkweigher ebook
Delivering consistent quality products is essential to protect your brand and your bottom line. That means knowing that the weight of a packaged product being shipped out the door matches the weight on the label. Do you have the right equipment for providing reliable weight control? To find out how checkweighers work, and which checkweigher system will work best for you, read A Practical Guide to Checkweighers and Checkweighing.
Download the free eBook
Video spotlight