Chemical identification solutions for detecting chemical weapons, agents and toxic industrial materials
Identify unknown chemical threats in seconds
Rapidly identify chemical warfare agents (CWAs), toxic industrial chemicals (TICs), toxic industrial materials (TIMs). Thermo Scientific chemical identification analyzers are compact and portable, and use multiple technologies to deliver a fast, confident response in the field. Easily decontaminated and certified to MIL-STD 810G standards for ruggedness, our instruments can be used  by military personnel right in the hazard zone, minimizing risk of contamination and exposure.
Contact a sales representative
Featured products to detect chemical threats
Gemini™ Analyzer
FirstDefender™ RMX Handheld Chemical Identification
TruDefender™ FTX/FTXi Handheld FTIR for Chemical Identification
Gain a significant tactical advantage in the field when you need fast identification of a broad range of unknown chemicals and explosives. The Thermo Scientific Gemini Analyzer helps military personnel make life-saving decisions using FTIR and Raman spectroscopy in a single lightweight, handheld instrument.
Increase flexibility and safety in incident response situations that call for rapid, accurate chemical identification with the Thermo Scientific FirstDefender RMX Handheld Chemical Identification Analyzer. The FirstDefender analyzer can be used as a handheld instrument or integrated onto a tactical robot to quickly analyze potentially hazardous materials.
Quickly identify explosives or hazardous materials with Thermo Scientific TruDefender handheld analyzers. TruDefender FTX and FTXi instruments harness the power of FTIR spectroscopy to provide fast, actionable data to mitigate potential threats and protect hazmat teams and the community.
Video spotlight: Chemical identification solutions