Search
Primary XPS region: B1s
Overlapping regions: Si2s plasmon, P2s, Zr3d
Binding energies of common chemical states:
| Chemical state | Binding energy |
|---|---|
| B elemental | 187.2 eV |
| ZrB2 | 187.8 eV |
| B sub-oxide[1] | 188.6 eV |
| Li2B4O7 | 192.5 eV |
Charge referenced to adventitious C1s peak at 284.8eV.
General comments
References
About this element
Symbol: B
Date of discovery: 1808
Name origin: Arabic buraq
Appearance: brownish
Discoverer: Sir Davy, et al.
Obtained from: kernite
Melting point: 2349 K
Boiling point: 4200 K
Density[kg/m3]: 2460
Molar volume: 4.39 × 10-6 m3/mol
Protons/electrons: 5
Neutrons: 6
Shell structure: 2,3
Electron configuration: [He]2s22p1
Oxidation state: 3
Crystal structure: rhombohedral
Although compounds of boron have been known for thousands of years, the element itself was not isolated until Sir Humphry Davy, Gay-Lussac, and L.J. Thenard isolated it to about 50% purity in 1808. Jöns Jacob Berzelius later identified boron as an element in 1824 and American chemist, W. Weintraub, produced pure boron for the first time in 1909. Boron is not found in its elemental form in nature, but is found in compounds. Compounds of boron have many applications: they are used in making insulating fiberglass, sodium perborate bleach, pyrotechnic flares, and making high-strength, lightweight materials primarily important for advanced aerospace structures.
Products
最適なシステム性能をお届けするため、当社は国際的なネットワークで、分野ごとのサービスエキスパート、テクニカルサポート、正規交換部品などを提供しています。