Organism TypeMouseThese cells are epithelial cells. The F9 cells differentiate if stimulated with retinoic acid and dibutyryl cyclic AMP (cAMP). Differentiating cells synthesize plasminogen activator, laminin and type IV collagen.
TissuesTestes
PhenotypeAdherent
Primaryno
ApplicationTransient Transfection