Connexin 26 / GJB2 Antibody (335800) in IHC
Immunofluorescence analysis of connexin 26 was performed on sections of adult mouse liver. Tissue sections on slides were probed for 24 h at 4°C in a humidified chamber with a mouse monoclonal anti-Cx26 (Cat. No. 33-5800), at an antibody concentration of 1-2 µg/ml diluted in 50 mM Tris-HCl, pH 7.4, containing 1.5% NaCl, 0.3% Triton X-100 (TBST) and 4% normal goat serum. After overnight incubation, sections were washed extensively for 1 h in TBST, and detection of primary antibody was performed for 1.5 h at room temperature with AlexaFluor-488-conjugated donkey anti-mouse diluted 1:600 in TBST. Sections were then was in TBST, then in TBS (without triton) and then coversliped with anti-fade medium. Images were taken on a Zeiss Z2 scanning microscope at x40 objective magnification, and show immunofluorescence labelling of Cx26 localized at gap junctions between liver hepatocytes. Data courtesy of Dr. James Nagy's lab.
Immunofluorescence analysis of connexin 26 was performed on sections of adult mouse liver. Tissue sections on slides were probed for 24 h at 4°C in a humidified chamber with a mouse monoclonal anti-Cx26 (Cat. No. 33-5800), at an antibody concentration of 1-2 µg/ml diluted in 50 mM Tris-HCl, pH 7.4, containing 1.5% NaCl, 0.3% Triton X-100 (TBST) and 4% normal goat serum. After overnight incubation, sections were washed extensively for 1 h in TBST, and detection of primary antibody was performed for 1.5 h at room temperature with AlexaFluor-488-conjugated donkey anti-mouse diluted 1:600 in TBST. Sections were then was in TBST, then in TBS (without triton) and then coversliped with anti-fade medium. Images were taken on a Zeiss Z2 scanning microscope at x40 objective magnification, and show immunofluorescence labelling of Cx26 localized at gap junctions between liver hepatocytes. Data courtesy of Dr. James Nagy's lab.
Related Products
Related Images