Search
IP-MS enrichment of TP53 (LFQ intensity): TP53 was enriched 318-fold from BT-549 lysate compared to background proteins, using the optimized IP-MS workflow with Pierce MS-Compatible Magnetic IP Kit protein A/G (Part No. 90409) and TP53 antibody (Part No. MA5-12557). See more information on IP-MS verification of antibody selectivity. Red color identifies the antibody's intended target, and blue color identifies direct interactors of that target protein.