UBE2M Antibody (PA5-42283) in WB
Western blot analysis of mouse brain cells using an anti-UBE2M polyclonal antibody (Product # PA5-42283). Primary Antibody Dilution: 2 µg/ml; Secondary Antibody: IRDye 800; Secondary Antibody Dilution: 1:20,000.; LANE 2) mouse brain extracts (80µg).
Western blot analysis of mouse brain cells using an anti-UBE2M polyclonal antibody (Product # PA5-42283). Primary Antibody Dilution: 2 µg/ml; Secondary Antibody: IRDye 800; Secondary Antibody Dilution: 1:20,000.; LANE 2) mouse brain extracts (80µg).
Related Products
Related Images