kits and reagents for sample preparation
Store and prep your sample with confidence
Preserving the integrity of your samples is critical to the success of every biobanking project. Biological samples must be prepared for a range of different research objectives and downstream analysis. Our solutions support the stabilization and preparation of a wide range of sample types offering you accurate, reliable performance that you can trust. 
Recover DNA and RNA from the same formaldehyde- or paraformaldehyde-fixed paraffin-embedded (FFPE) tissue sample. The DNA and RNA are recovered in separate eluates, and both are compatible with a broad range of applications, including real-time PCR and next-generation sequencing.. Common nucleic acid isolation methods require you to choose which nucleic acid to recover from a given sample, with loss of the other nucleic acid (and any information that may have resulted from its analysis). Alternatively, the DNA and RNA are co-purified, or the sample is split in half so that each can be purified.
Recover DNA and RNA from the same formaldehyde- or paraformaldehyde-fixed paraffin-embedded (FFPE) tissue sample. The DNA and RNA are recovered in separate eluates, and both are compatible with a broad range of applications, including real-time PCR and next-generation sequencing.. Common nucleic acid isolation methods require you to choose which nucleic acid to recover from a given sample, with loss of the other nucleic acid (and any information that may have resulted from its analysis). Alternatively, the DNA and RNA are co-purified, or the sample is split in half so that each can be purified.