cold storage equipment
Ultra-low and cryogenic freezers designed to protect critical samples
Our cold storage equipment portfolio offers a wide range of ultra-low upright and chest freezers, as well as LN2 auto- and manual-fill freezers to maximize sample storage capacity within the footprint of the biobank. Our new TSX Series of ultra-low freezers have been designed to offer an energy-efficient, solution without compromising the importance of sample protection.
Featured cold storage equipment for biobanking
Thermo Scientific™ CryoExtra™ auto-fill LN2 storage systems are designed for both liquid and vapor phase storage. Four capacities allow you to store 19,500 up to 93,000 2mL vials.
cryoextra high-efficiency cryogenic storage systems