Search
| Materials Science | Semiconductors | Life Sciences | |||||||||
|---|---|---|---|---|---|---|---|---|---|---|---|
| Features | Talos F200x | Talos F200i | Talos F200S | Talos F200E | Talos F200C | Talos 12 | |||||
| Pole piece | X-TWIN | X-TWIN | X-TWIN | C-TWIN | |||||||
| Gun type | X-FEG | X-CFEG | S-FEG | X-FEG | X-CFEG | S-FEG | X-FEG | X-FEG | LaB6/W | ||
| Gun brightness at 200 kV [A/cm2/sr] |
1.8·10⁹ | 2.4·10⁹ | 4.0-108 | 1.8-109 | 2.4·10⁹ | 4.0-108 | 1.8-109 | 1.8-109 | N/A | ||
| EDS type/area | In-column |
Side-entry 30 mm2 |
Side-entry 100 mm2 |
Side-entry Symmetrical Dual-X (200 mm2) |
In-column 2SDD Super-X (60 mm2) |
In-column 4SDD Super-X (120mm²) |
Side-entry Symmetrical Dual-X (200mm²) |
Side-entry 30/100 mm2 |
|||
| EDS net solid angle [sr] |
0.9 | 0.19 | 1.28 | 1.65# | 0.45 | 0.90 | 1.65 | 0.18/0.80 | |||
| Velox absorption correction for most accurante quantification |
Yes | Yes | Yes | Yes | Yes | Yes | Yes | Yes | |||
| Automated/ unattended 3D EDS |
Yes | No | No | Yes | No | Yes | No | No | |||
| Maps / Automated Particale Workflow |
(S)TEM and EDS | (S)TEM and EDS |
(S)TEM and EDS |
(S)TEM and EDS |
(S)TEM and EDS |
(S)TEM and EDS |
|||||
| Cleanest EDS; Fiori P/B (on system) |
Excellent | Great | Great | Excellent | Great | Good | |||||
| HRTEM info limit |
0.12 | 0.11 | 0.12 | 0.11 | 0.12 | 0.12 | 0.18 | N/A | |||
| HRTEM line resolution |
0.10 | 0.10 | 0.10 | 0.10 | 0.10 | 0.10 | 0.20 | ||||
| HRSTEM [nm] | 0.16 | 0.14 |
0.16 | 0.14 (with >100 pA) |
0.16 | 0.16 | 0.20 | 1 | |||
| Align Genie | Yes | Yes | Yes | Yes | Yes | - | |||||
| EELS resolution | 0.8 eV | 0.3 eV | 0.8 eV | 0.3 eV | 0.8 eV | 0.8 eV | 0.3 eV | 0.8 eV | ~1.5 eV | ||
| CrystalPack |
Yes | Yes | Yes | Yes | Yes | Yes | |||||
| iDPC | Yes | Yes | Yes | Yes | Yes | Yes | |||||
| AutoSTEM | Yes | Yes | Yes | Yes | Yes | No | |||||
| Enclosure | Optimized,incl. touch screen |
Optimized, incl. touch screen | Compact | Optimized, incl. touch screen |
Optimized, incl. touch screen |
Optimized, incl. touch screen |
- | ||||
| Cryo quality | Great | Great | N/A | Excellent | |||||||
| TEM contrast | N/A | Excellent | |||||||||
| Phase plate | No | No | No | No | Yes | ||||||
| Semiconductor standard compliant |
Optional | N/A | Yes | N/A | |||||||
| TEM image linear distortion specification |
N/A | N/A | Yes |
N/A | |||||||
| TEM image linear variation specification |
N/A | N/A | Optional |
N/A | |||||||
#=the geometric angle is 2.55
For Research Use Only. Not for use in diagnostic procedures.