Custom siRNA and miRNA products provide a tailored solution for researchers looking to implement RNA interference in their studies. We offer high-quality synthesis and flexible customization options to help ensure researchers achieve precise and efficient gene silencing results.
Start your project now
Custom RNA modification services
Whether you need a modified predesign reagent, have your own modified sequence, or are targeting a different species, our team is here to help! We provide a range of custom siRNA modification services to cater to your distinct requirements.
Our highly trained scientists can custom-synthesize almost any single-stranded RNA, siRNA or miRNA you may need. Whether you require a single molecule or thousands, your project will receive meticulous handling and quality control throughout the entire process.
Reasons for modifying siRNA
  • Improved stability and efficacy
  • Reduced toxicity and immunogenicity
  • Monitor or facilitate delivery
Types and applications of RNA modifications
Types of RNA modificationsModificationReason for modification
Dye labelsAlexa Fluor dyes
Cy3
Cy5
6’ FAM
Monitor delivery
Sugar modificationsO-Methyl (2’ OMe)
Fluoro (2’ F)
O-methoxy ethyl (2’ MOE)
Locked nucleic acid (LNA)
Prevent nuclease activity
Provide stability
Reduce immunogenicity
Backbone modificationsPhosphorothioate (PS)
Methyl Phosphonate (MP)
Methyl Phosphorothioate
Increase nuclease resistance
Reactive groupsBiotin
Phosphate
Thiol
Amino modifier
DBCO
Azide
Enable downstream chemical reactions (i.e., ligand binding, enzymatic ligation, molecule conjugation)
Delivery moietiesGalNAc
Fatty acid
Facilitate delivery
Don’t see your modification? Complete a quote request to learn more about how we may be able to support your custom needs.
Request a quote
Custom siRNA and miRNA design and ordering tools
By using our design tools, you can create custom sequences for any organism. The designers can create custom duplexes based on a cDNA sequence, Entrez ID or GenBank accession number, or by entering your existing sequence.
Custom designed siRNA duplexes provide
  • Designs using our proprietary Silencer Select algorithm for creating highly potent and specific designs
  • All of the advantages of the chosen product chemistry
  • Matched controls for any organism
Products supportedCustomization needOrdering tool
Silencer Select siRNA
Silencer siRNA
Ambion In Vivo siRNA
  • Transcript specific targeted siRNA
  • siRNA for species other than human, mouse, and rat
  • Custom siRNA sequences
GeneAssist siRNA
mirVana miRNA inhibitors
mirVana miRNA mimics
Ambion Anti-miR miRNA Inhibitors
Ambion Pre-miR miRNA Precursors
  • Custom miRNA sequences not available in our predesigned catalog
GeneAssist miRNA
Stealth RNAi siRNA
shRNA
  • Custom Stealth RNAi siRNA sequence
  • Convert an existing siRNA sequence to Stealth RNAi siRNA
  • Designing shRNA sequences for specific genes
  • Converting siRNA to shRNA
RNAi Designer
Single-stranded RNA
  • Synthesis of up to 50 nucleotides
Custom RNA Oligos
Custom modified RNAi order process
Custom RNAi: Complete our custom quote request, and a member of our team will reach out to confirm your project requirements. After gathering all necessary details, you will receive a quote and order instructions. We have a dedicated RNAi support team to assist in order placement and provide any details throughout the project, as needed.
Turnaround time (TAT): The turnaround time for custom RNAi reagents depends on the scale and complexity of the order. For smaller scale synthesis ordered through our online tool, we can usually produce custom sequences within 4–10 business days. More complex and highly modified sequences require assessment by our manufacturing team and typically ship in 20–25 business days.
Need assistance? If you have any questions or need help, contact our project managers at RNAiSupport@thermofisher.com.
High quality siRNA synthesis
We synthesize and purify each siRNA in state-of-the-art facilities to meet the highest quality standards. All offline custom synthesis orders have a minimum requirement of delivering 50 nmol HPLC-purified material. As part of our rigorous quality control procedures, each RNA oligonucleotide is analyzed by MALDI-TOF mass spectrometry and/or analytical HPLC. To offer the utmost in quality, we also assess each annealed siRNA by gel electrophoresis to confirm that the strands annealed properly. The result is premium quality siRNA that is purified and ready to use.
Custom siRNA and miRNA libraries
For those requiring custom libraries, we provide plated sets of siRNAs and miRNA mimics and inhibitors designed to target your list of genes.
See our flyer for available library information
Contact us today for a quote
For Research Use Only. Not for use in diagnostic procedures.
Hero banner dark gradient overlay