Search
Solving materials science challenges relies on nanoscale characterization, combining data from various techniques. Thermo Fisher Scientific provides instruments and advanced software for a range of analytical techniques, including scanning electron microscopy, focused ion beam milling, transmission electron microscopy, X-ray photoemission spectroscopy, energy-dispersive spectroscopy, electron backscatter diffraction, and electron energy loss spectroscopy.
With smaller overall size, desktop SEM instruments enhance ease of use and democratize SEM technology. While traditional floor model SEM instruments can require a specialized room or facility, desktop instruments are far more robust. Some researchers have even taken their desktop SEMs on the road, providing on-site electron microscopy analysis via mobile laboratories. Desktop SEMs often provide the shortest time to data in failure analysis of electrolyzer or fuel cell components and characterize contamination with built-in compositional analysis.
Floor model SEMs offer the flexibility and versatility to meet a wide range of academic and industrial needs: support for large and heavy samples, a very wide range of accessories, excellent imaging quality for the most challenging materials or the smallest details, and dynamic experimentation. Modern SEMs can easily image catalyst nanoparticles and beam-sensitive polymer membranes, and they can also characterize all materials in an electrolyzer or fuel cell.
Thermo Fisher Scientific is the industry leader in FIB-SEM technology with more than 30 years of experience with our Thermo Scientific DualBeam Instruments. The technology’s ability to make site-specific cross sections, acquire 3D datasets, and prepare TEM lamellae is crucial for electrolyzer and fuel cell R&D.
TEM instruments are capable of unparalleled resolution and routinely collect sub-nanometer scale (if not atomic resolution) images. With electrochemical processes in electrolyzers and fuel cells that depend on thin layers, small particles, and contamination in such features, a TEM is an indispensable tool for research and development.
X-ray photoelectron spectroscopy instruments are the standard tools for surface characterization, providing information about the composition and chemical state of materials in the top few nanometers of a surface, with an option to dig deeper into the surface as well. XPS is crucial for the characterization and development of catalyst materials.
For Research Use Only. Not for use in diagnostic procedures.