Search
Primary XPS region: Cs3d
Overlapping regions: N/A
Binding energies of common chemical states:
| Chemical state | Binding energy Cs3d5/2 |
|---|---|
| Csl | 724 eV |
N/A
Symbol: Cs
Date of discovery: 1860
Name origin: Latin caesius
Appearance: silver
Discoverer: Fustov Kirchoff
Obtained from: pollucite, lepidolite
Melting point: 28.5 K
Boiling point: 678.4 K
Density[kg/m3]: 1.873
Molar volume: 70.94 × 10-6 m3/mol
Protons/Electrons: 55
Neutrons: 78
Shell structure: 2,8,18,18,8,1
Electron configuration: [Xe]6s1
Oxidation state: 1
Crystal structure: cubic
Cesium, also spelled caesium, is a silvery gold metal that is liquid at or around room temperature. This element’s name is derived from the Latin caesius, meaning sky blue. Cesium reacts explosively with water. Cesium hydroxide is a very strong base that can rapidly etch the surface of glass. The most widespread use of cesium is in drilling fluids for the oil industry which mitigates the necessity of toxic suspended solids in drilling fluids. Cesium is also used in atomic clocks, in particular its isotope of atomic weight 133 (Cs-133).
Products
最適なシステム性能をお届けするため、当社は国際的なネットワークで、分野ごとのサービスエキスパート、テクニカルサポート、正規交換部品などを提供しています。