Search
Join the conversation
Primary XPS region: Er4d
Overlapping regions: Si2s plasmons
Binding energies of common chemical states: N/A
| Chemical state | Binding energy Er4d |
|---|---|
| Er metal | 167.5 eV |
| Er native oxide | ~170 eV |
Native oxide charge referenced to adventitious C1s peak at 284.8 eV.
N/A
Symbol: Er
Date of discovery: 1842
Name origin: Ytterby, Sweden
Appearance: grayish
Discoverer: Carl Gustof Mosander
Obtained from: xenotime, euxenite
Melting point: 1770 K
Boiling point: 3141 K
Density[kg/m3]: 9066
Molar volume: 18.46 × 10-6 m3/mol
Protons/Electrons: 68
Neutrons: 99
Shell structure: 2,8,18,30,8,2
Electron configuration: [Xe]6s24f12
Oxidation state: 3
Crystal structure: hexagonal
Erbium, along with other rare elements, is in the mineral gadolinite from Ytterby in Sweden. It is used as a photographic filter and also as a neutron absorber in nuclear technology. Because of its oxide’s pink color, Erbium is sometimes used as a glass and porcelain enamel glaze colorant-yielding glass that is used in sunglasses and inexpensive jewelry. Metallic erbium’s dust presents a fire and explosion hazard. This element gives characteristic sharp absorption spectra in the near infrared, visible light, and ultraviolet regions. Erbium plays no known biological role and is never found free in nature.
최적의 시스템 성능을 보장하기 위해 세계 수준의 현장 서비스 전문가 네트워크, 기술 지원 및 인증된 예비 부품을 제공해드립니다.