Search
Natural Mouse Tau, from mouse brain homogenate, was serially diluted in Standard Diluent Buffer. The optical density of each dilution was plotted against the standard curve. Parallelism between the natural and recombinant protein was demonstrated by the figure below and indicated that the standard accurately reflects natural Mouse Tau content in samples.