Search
Optimize your experiments to get the best results. We’ve compiled a detailed knowledge base of the top tips and tricks to meet your research needs.
View the relevant questions below:
Having problems with your experiment? Visit our
Unfortunately, GeneMapper® Software v.4.0 is not Windows® 7 compatible. Please click here for the GeneMapper® Software v.4.0 datasheet.
The computer requirements are as follows:
*Hard drive requirements will vary depending on how much space is allocated to the Oracle Database. Having a large hard drive will allow for database expansion should the database fill up.
For the operating system, GeneMapper® Software v.4.0 requires Windows® XP Professional, service pack 1 or 2, or Windows® 2000, service pack 4. Please click here for the GeneMapper ®Software v.4.0 datasheet.
Unfortunately, GeneMapper® Software v.4.1 is not compatible with Windows® 7. Please click here for the GeneMapper® Software v.4.1 datasheet.
Please see the computer requirements below:
Recommended CPU: Core 2 Duo E8400/3.0 GHz
*Hard drive requirements will vary depending on how much space is allocated to the Oracle Database. Having a large hard drive will allow for database expansion should the database fill up.
For the operating system, GeneMapper® Software v.4.1 requires Windows® XP Professional, service pack 2 or 3, or Windows Vista™ (32-bit Business Edition), service pack 1. For more information, please click here for the GeneMapper® Software v.4.1 datasheet.
GeneMapper® Software v4.1 is compatible with the following data files:
Please note, upgrading the Data Collection Software on the 3130 and 3730 series instruments to v.3.1 for co-installation is required. For more information, please click here for the GeneMapper® Software v.4.1 datasheet.
GeneMapper® Software 5 has been extensively tested on Windows® 7 SP1 (32-bit version). While there are some indications that software will run normally on the 64-bit version of Windows® 7 SP1, the 64-bit configuration is not officially supported. It is strongly recommended that this software be run on Windows® 7 SP1 (32-bit). For a datasheet on GeneMapper® Software 5, please click here.
If installing on the instrument computer, the update will require a new computer and upgrading of the Data Collection Software as well.
GeneMapper® Software 5 is compatible with the following files:
Please click here to get the datasheet for GeneMapper® Software 5.
Please see the computer requirements below for GeneMapper® Software 5:
Recommended CPU: Core 2 Duo E8400/3.0 GHz
For the GeneMapper® Software 5 datasheet, please click here.
For Research Use Only. Not for use in diagnostic procedures.