LanthaScreen Kinase Activity Assay Selection Table
Our LanthaScreen kinase activity assay toolbox of 'mix and match' assay reagents is designed to enable rapid assay configuration. Our portfolio of phospho-specific antibodies, matched fluorescent peptide substrates and in-house produced kinases have been tested in >200 different kinase assays to date. See below for a list of kinases that have been tested in the LanthaScreen Technology.
Additional antibodies from the table below may be available with Europium labels, contact us at discoverysciences@thermofisher.com, for more information.
Search LanthaScreen Kinase Table alphabetically
All Kinase - Alphabetical List:
A - B - C - D - E - F - G - H - I - J - K - L - M - N - O - P - Q - R - S - T - U - V - W - X - Y - Z
Kinase Kinase Alias(es) LanthaScreen Tb Activity Assay LanthaScreen Eu Activity Assay
Antibody Substrate Antibody  Substrate
A BL1 ABL, c-ABL Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 AlexaFluor  647-Poly GT
ABL1E2 55K ABL, c-ABL Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
ABL1 G250E ABL, c-ABL Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
ABL1 T315I ABL, c-ABL Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
ABL1 Y253F ABL, c-ABL Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
ABL2 Arg Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
AKT1 PKB alpha, RAC, PRKBA Tb-pCrosstide Fluorescein-Crosstide    
AKT2 PKB beta, PRKBB,RAC beta Tb-pCrosstide Fluorescein-Crosstide    
AKT3 PKB gamma, PRKBG,RAC-gamma Tb-pCrosstide Fluorescein-Crosstide    
ALK   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
AMPK (A1/B1/G1) PRKAA1/ PRKAB1/ PRKAG1 Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
AMPK(A 2/B1/G1) PRKAA2/PRKAB1/PRKAG1 Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
AURKA Aurora A, AIK, AURA, STK6 Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
AURKB Aurora B, AIK2, ARK2 Tb-pSer (pPKC) Fluorescein-PKC Inquire Inquire
AXL UFO Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
BLK MGC10442 Tb-PY- 20 Fluorescein-Poly GT    
BMX ETK, PSCTK3, PSCTK2 Tb-PY- 20 Fluorescein-Poly GT    
BRAF BRAF1, RAFB1 Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1 Inquire Inquire
BRAF V599E BRAF1, RAFB1 Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1 Inquire Inquire
BTK ATK, XLA Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
CAMK1   Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
CAMK1D CaMKI delta, CLKiK Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
CAMK2A CaMKII alpha, CAMKA, KIAA0968 Tb-pCrosstide Fluorescein-Crosstide    
CAMK2B CaMKII beta, CAMKB, CAM2, CAMK2 Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
CAMK2D CaMKII delta Tb-pCr osstide Fluorescein-Crosstide    
CAMK4 CaMKIV, CAMK-GR, MGC36771 Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
CHEK1 CHK1 Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
CHEK2 CHK2, CDS1 Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
CHUK IKK-alpha, Chuk1, IKK1 Tb-IkB a pSer32 Fluorescein-IKK    
CSF1R FMS, CSFR, c-FMS, CD115 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
CSK   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
DDR2 TKT, NTRKR3 Tb-PY- 20 Fluorescein-Poly GT    
DNA-PK PRKDC Tb-pho spho-p53 [pSer15] Fluorescein-p53 Peptide Inquire Inquire
DYRK4   Tb-pSe r (pPKC) Fluorescein-PKC    
EGFR ErbB1 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
EGFR L858R ErbB1 L858R Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
EGFR L861Q ErbB1 L861Q Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
EGFR T790M ErbB1 T790M Tb-PY- 20 Fluorescein-Poly GT    
EIF2AK 1 HRI Tb-peI F2 alpha [pSer52] GFP-eI F2a    
EIF2AK 2 MGC126524, PKR, PRKR Tb-peI F2 alpha [pSer52] GFP-eI F2a    
EIF2AK 3 PERK Tb-peI F2 alpha [pSer52] GFP-eI F2a    
EPHA1 EPH Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
EPHA2 ECK Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
EPHA3 HEK, HEK4 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
EPHA4 HEK8 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
EPHA5 CEK7, EHK1, HEK7, TYRO4 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
EPHA7 HEK11, EHK3 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
EPHA8 HEK3, EEK Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
EPHB1 HEK6 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
EPHB2 HEK5 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
EPHB3 HEK2, TYRO6 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
EPHB4 TYRO11 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
ERBB2 HER2, NEU Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
ERBB4 HER4 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
FER TYK3 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
FES Fps Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
FGFR1 H2, H3, H4 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
FGFR2 CEK3, TY14, BEK Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
FGFR3 CEK2, JTK4 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
FGFR3 K650E ACH, CEK2 Tb-PY- 20 Fluorescein-Poly GT    
FGFR4 JTK2 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
FGR c-fgr, p55c-fgr, SRC2 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
FLT1 VEGFR1, FLT Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
FLT3 FLK2, STK1, CD135 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
FLT3 D835Y FLK2, STK1, CD135 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
FLT4 VEGFR3, PCL Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
FRAP1 mTOR, FRAP, FRAP2, TOR, RAFT1 Tb-p4E -BP1 [pThr46] GFP-4E -BP1 Inquire Inquire
FRAP1 mTOR, FRAP, FRAP2, TOR, RAFT1     Inquire Inquire
FRK PTK5, GTK, RAK Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
FYN SLK, SYN, FynT Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
GRK5 GPRK5 Tb-IkB a pSer32 Fluorescein-IKK    
GRK6 GPRK6 Tb-IkB a pSer32 Fluorescein-IKK    
HCK JTK9 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
IGF1R JTK13, CD221 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
IKBKB IKK beta, NFKBIKB, IKK2 Tb-IkB a pSer32 Fluorescein-IKK    
IKBKE IKK epsilon, IKKE, IKKI Tb-IkB a pSer32 Fluorescein-IKK    
INSR   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
INSRR IRR Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
ITK EMT, LYK, PSCTK2 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
JAK1 JAK1A, JAK1B Tb-pST AT1 [pTyr701] GFP-ST AT1    
JAK2   Tb-pST AT1 [pTyr701] GFP-ST AT1    
JAK2 JH1 JH2   Tb-pST AT1 [pTyr701] GFP-ST AT1    
JAK2 JH1 JH2 V617F   Tb-pST AT1 [pTyr701] GFP-ST AT1    
JAK3 LJAK, JAKL Tb-pST AT1 [pTyr701] GFP-ST AT1    
KDR VEGFR2, VEGFR, FLK1 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
KIT cKit, PBT, SCFR, CD117 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
KIT T670I cKit, PBT, SCFR, CD117 Tb-PY- 20 Fluorescein-Poly GT    
KIT V654A cKit, PBT, SCFR, CD117 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
LCK   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
LRRK2 AURA17, PARK8, RIPK7, ROCO2 Tb-ERM (pLRRKtide) Fluorescein-ERM (LRRKtide) Inquire Inquire
LRRK2 G2019S AURA17, PARK8, RIPK7, ROCO2 Tb-ERM (pLRRKtide) Fluorescein-ERM (LRRKtide) Inquire Inquire
LRRK2 I2020T AURA17, PARK8, RIPK7, ROCO2 Tb-ERM (pLRRKtide) Fluorescein-ERM (LRRKtide) Inquire Inquire
LRRK2 R1441C AURA17, PARK8, RIPK7, ROCO2 Tb-ERM (pLRRKtide) Fluorescein-ERM (LRRKtide) Inquire Inquire
LTK TYK1 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
LYNA   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
LYNB   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
MAP3K1 0 MLK2, MST Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1    
MAP3K1 1 MLK3, SPRK Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1 Inquire Inquire
MAP3K2 MEKK2, MEKK2B Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1 Inquire Inquire
MAP3K3 MEKK3, MAPKKK3 Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1 Inquire Inquire
MAP3K5 ASK1, MAPKKK5 Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1 Inquire Inquire
MAP3K7 / MAP3K7 IP1 TAK1/TAB1 Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1 Inquire Inquire
MAP3K8 COT1, EST, ESTF Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1 Inquire Inquire
MAP3K9 MLK1, PRKE1 Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1 Inquire Inquire
MAP4K2 GCK, BL44 Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire
MAP4K4 HGK, NIK Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire
MAP4K5 KHS, GCKR Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire
MAPK1 ERK2, ERK Tb-pAT F2 [pThr71] GFP-AT F2    
MAPK10 JNK3 Tb-pAT F2 [pThr71] GFP-AT F2    
MAPK11 p38 beta, P38b, P382, SAPK2 Tb-pAT F2 [pThr71] GFP-AT F2    
MAPK12 p38 gamma, ERK6, SAPK3 Tb-pAT F2 [pThr71] GFP-AT F2    
MAPK13 p38 delta, SAPK4 Tb-pAT F2 [pThr71] GFP-AT F2    
MAPK14 p38 alpha, p38, Mxi2, SAPK2A Tb-pAT F2 [pThr71] GFP-AT F2    
MAPK3 ERK1, P44ERK1, PRKM3 Tb-pAT F2 [pThr71] GFP-AT F2    
MAPK8 JNK1, JNK, SAPK1 Tb-pAT F2 [pThr71] GFP-AT F2    
MAPK9 JNK2 Tb-pAT F2 [pThr71] GFP-AT F2    
MAPKAP K2 MK2 Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
MAPKAP K3 3PK, MAPKAP3 Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
MARK4 KIAA1860 Tb-pSe r (pPKC) Fluorescein-PKC    
MATK Hyl, CTK Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
MERTK cMER. MER Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
MET cMet, HGFR, RCCP2 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
MET M1250T cMet, HGFR, RCCP2 Tb-PY- 20 Fluorescein-Poly GT    
MINK1 MINK, B55, ZC3 Tb-pSe r (pPKC) Fluore scein-PKC    
MST1R RON, CDw136 Tb-PY- 20 Fluorescein-Poly GT    
MST4   Tb-pSe r (pPKC) Fluorescein-PKC    
MUSK   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
NEK2 NLK1, HsPK21 Tb-pSe r (pPKC) Fluorescein-PKC    
NLK FLJ21033, DKFZp761G1211 Tb-pc- Jun [pSer73] GFP-c- Jun    
NTRK1 TRKA, MTK, TRK Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
NTRK2 TRKB Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
NTRK3 TRKC Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
PAK4   Tb-pCr osstide Fluorescein-Crosstide    
PAK7 PAK5 Tb-pCr osstide Fluorescein-Crosstide    
PASK   Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire  
PDGFRA PDGFR alpha, CD140A, PDGFR2 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
PDGFRA D842V PDGFR alpha, CD140A, PDGFR2 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
PDGFRA T674I PDGFR alpha, CD140A, PDGFR2 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
PDGFRA V561D PDGFR alpha, CD140A, PDGFR2 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
PDGFRB PDGFR beta, JTK12, CD140B, PDGFR1 Tb-PY- 20 Fluorescein-Poly GT    
PHKG1   Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire
PIM1   Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
PIM2   Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
PKN1 PRK1 Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
PRKACA PKA Tb-pCr osstide Fluorescein-Crosstide    
PRKCA PKC alpha Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire
PRKCB1 PKC beta I Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire
PRKCB2 PKC beta II Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire
PRKCD PKC delta Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire
PRKCE PKC epsilon Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire
PRKCG PKC gamma Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire
PRKCH PKC eta Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire
PRKCI PKC iota Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire
PRKCN PKD3, PRKG3, EPK2, PKC nu Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
PRKCQ PKC theta Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire
PRKCZ PKC zeta Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire
PRKD1 PKD, PKC mu Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
PRKD2 PKD2, HSPC187 Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
PRKG1 PGK, CGK1, FLJ36117 Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
PRKG2 PKG2, PRKGR2, cGKII Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
PRKX PKX1 Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire
PTK2 FAK, FADK, FAK1 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
PTK2B FAK2, CAKB, PYK2 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
PTK6 Brk Tb-PY- 20 Fluorescein-Poly GT    
RAF1 (cRaf) Y340D Y341D   Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1    
RET   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
RET V804L   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
RET Y791F   Tb-PY- 20 Fluorescein-Poly GT    
ROCK1   Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire
ROCK2 KIAA0619 Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire
ROS1 ROS, MCF3 Tb-PY- 20 Fluorescein-Poly GT    
RPS6KA 1 RSK1, HU-1 Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire
RPS6KA 2 RSK3, HU-2 Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire
RPS6KA 3 RSK2, HU-3 Tb-pCr osstide Fluorescein-Crosstide Inquire Inquire
RPS6KA 4 MSK2, RSK-B Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
RPS6KA 5 MSK1, RLPK Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
RPS6KB 1 p70S6K, STK14A, S6K Tb-pCr osstide Fluorescein-Crosstide    
SGK SGK1 Tb-pSe r (pPKC) Fluorescein-PKC    
SGK2   Tb-pSe r (pPKC) Fluorescein-PKC    
SGKL SGK3 Tb-pSe r (pPKC) Fluorescein-PKC    
SRC ASV Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
SRCN1   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
SRMS SRM Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
STK22D TSSK1, FKSG81, SPOGA4 Tb-pCREB [pSer133] Fluorescein-CREBtide Inquire Inquire
STK24 MST3, STK3 Tb-pSe r (pPKC) Fluorescein-PKC    
STK25 YSK1, SOK1 Tb-pSe r (pPKC) Fluorescein-PKC    
SYK   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
TAOK2 TAO1, PSK, TAO2, KIAA0881 Tb-pSe r (pPKC) Fluorescein-PKC    
TAOK3 JIK Tb-pCr osstide Fluorescein-Crosstide    
TBK1 NAK, T2K, FLJ11330 Tb-IkBa
Fluorescein-IKK
TEC PSCTK4 Tb-PY- 20 Fluorescein-Poly GT    
TEK Tie2, VMCM, CD202B Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
TGFBR1 ALK5 Tb-PY- 20 Fluorescein-Poly GT Inquire Inquire
TNK2 ACK Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
TTK MPS1L1 Tb-PY- 20 Fluorescein-Poly GT    
TXK   Tb-PY- 20 Fluorescein-Poly GT    
TYK2 JTK1 Tb-pST AT1 [pTyr701] GFP-ST AT1    
TYRO3 DKT, BRT, TIF Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
YES1 Yes, C-YES, P61-Yes Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
ZAP70   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT
Kinase Kinase Alias(es) LanthaScreen Tb Activity Assay LanthaScreen Eu Activity Assay  
Antibody Substrate Antibody  Substrate
Protocol
A BL1 ABL, c-ABL Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 AlexaFluor  647-Poly GT  
ABL1E2 55K ABL, c-ABL Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
ABL1 G250E ABL, c-ABL Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
ABL1 T315I ABL, c-ABL Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
ABL1 Y253F ABL, c-ABL Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
ABL2 Arg Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
AKT1 PKB alpha, RAC, PRKBA Tb-pCr osstide Fluorescein-Crosstide      
AKT2 PKB beta, PRKBB,RAC beta Tb-pCr osstide Fluorescein-Crosstide      
AKT3 PKB gamma, PRKBG,RAC-gamma Tb-pCr osstide Fluorescein-Crosstide      
ALK   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
AMPK (A1/B1/G1) PRKAA1/ PRKAB1/ PRKAG1 Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire Inquire  
AMPK(A 2/B1/G1) PRKAA2/PRKAB1/PRKAG1 Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire Inquire  
AURKA Aurora A, AIK, AURA, STK6 Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire Inquire  
AURKB Aurora B, AIK2, ARK2 Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire  
AXL UFO Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
BLK MGC10442 Tb-PY- 20 Fluorescein-Poly GT      
BMX ETK, PSCTK3, PSCTK2 Tb-PY- 20 Fluorescein-Poly GT      
BRAF BRAF1, RAFB1 Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1 Inquire Inquire  
BRAF V599E BRAF1, RAFB1 Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1 Inquire Inquire  
BTK ATK, XLA Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
CAMK1   Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire Inquire  
CAMK1D CaMKI delta, CLKiK Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire Inquire  
CAMK2A CaMKII alpha, CAMKA, KIAA0968 Tb-pCr osstide Fluorescein-Crosstide      
CAMK2B CaMKII beta, CAMKB, CAM2, CAMK2 Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire Inquire  
CAMK2D CaMKII delta Tb-pCr osstide Fluorescein-Crosstide      
CAMK4 CaMKIV, CAMK-GR, MGC36771 Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire Inquire  
CHEK1 CHK1 Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire Inquire  
CHEK2 CHK2, CDS1 Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire Inquire  
CHUK IKK-alpha, Chuk1, IKK1 Tb-IkB a pSer32 Fluorescein-IKK      
CSF1R FMS, CSFR, c-FMS, CD115 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
CSK   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
DDR2 TKT, NTRKR3 Tb-PY- 20 Fluorescein-Poly GT      
DNA-PK PRKDC Tb-pho spho-p53 [pSer15] Fluorescein-p53 Peptide Inquire Inquire  
DYRK4   Tb-pSe r (pPKC) Fluorescein-PKC      
EGFR ErbB1 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
EGFR L858R ErbB1 L858R Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
EGFR L861Q ErbB1 L861Q Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
EGFR T790M ErbB1 T790M Tb-PY- 20 Fluorescein-Poly GT      
EIF2AK 1 HRI Tb-peI F2 alpha [pSer52] GFP-eI F2a      
EIF2AK 2 MGC126524, PKR, PRKR Tb-peI F2 alpha [pSer52] GFP-eI F2a      
EIF2AK 3 PERK Tb-peI F2 alpha [pSer52] GFP-eI F2a      
EPHA1 EPH Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
EPHA2 ECK Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
EPHA3 HEK, HEK4 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
EPHA4 HEK8 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
EPHA5 CEK7, EHK1, HEK7, TYRO4 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
EPHA7 HEK11, EHK3 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
EPHA8 HEK3, EEK Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
EPHB1 HEK6 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
EPHB2 HEK5 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
EPHB3 HEK2, TYRO6 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
EPHB4 TYRO11 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
ERBB2 HER2, NEU Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
ERBB4 HER4 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
FER TYK3 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
FES Fps Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
FGFR1 H2, H3, H4 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
FGFR2 CEK3, TY14, BEK Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
FGFR3 CEK2, JTK4 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
FGFR3 K650E ACH, CEK2 Tb-PY- 20 Fluorescein-Poly GT      
FGFR4 JTK2 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
FGR c-fgr, p55c-fgr, SRC2 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
FLT1 VEGFR1, FLT Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
FLT3 FLK2, STK1, CD135 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
FLT3 D835Y FLK2, STK1, CD135 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
FLT4 VEGFR3, PCL Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
FRAP1 mTOR, FRAP, FRAP2, TOR, RAFT1 Tb-p4E -BP1 [pThr46] GFP-4E -BP1 Inquire Inquire  
FRAP1 mTOR, FRAP, FRAP2, TOR, RAFT1     Inquire Inquire  
FRK PTK5, GTK, RAK Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
FYN SLK, SYN, FynT Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
GRK5 GPRK5 Tb-IkB a pSer32 Fluorescein-IKK      
GRK6 GPRK6 Tb-IkB a pSer32 Fluorescein-IKK      
HCK JTK9 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
IGF1R JTK13, CD221 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
IKBKB IKK beta, NFKBIKB, IKK2 Tb-IkB a pSer32 Fluorescein-IKK      
IKBKE IKK epsilon, IKKE, IKKI Tb-IkB a pSer32 Fluorescein-IKK      
INSR   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
INSRR IRR Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
ITK EMT, LYK, PSCTK2 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
JAK1 JAK1A, JAK1B Tb-pST AT1 [pTyr701] GFP-ST AT1      
JAK2   Tb-pST AT1 [pTyr701] GFP-ST AT1      
JAK2 JH1 JH2   Tb-pST AT1 [pTyr701] GFP-ST AT1      
JAK2 JH1 JH2 V617F   Tb-pST AT1 [pTyr701] GFP-ST AT1      
JAK3 LJAK, JAKL Tb-pST AT1 [pTyr701] GFP-ST AT1      
KDR VEGFR2, VEGFR, FLK1 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
KIT cKit, PBT, SCFR, CD117 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
KIT T670I cKit, PBT, SCFR, CD117 Tb-PY- 20 Fluorescein-Poly GT      
KIT V654A cKit, PBT, SCFR, CD117 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
LCK   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
LRRK2 AURA17, PARK8, RIPK7, ROCO2 Tb-ERM (pLRRKtide) Fluorescein-ERM (LRRKtide) Inquire Inquire  
LRRK2 G2019S AURA17, PARK8, RIPK7, ROCO2 Tb-ERM (pLRRKtide) Fluorescein-ERM (LRRKtide) Inquire Inquire  
LRRK2 I2020T AURA17, PARK8, RIPK7, ROCO2 Tb-ERM (pLRRKtide) Fluorescein-ERM (LRRKtide) Inquire Inquire  
LRRK2 R1441C AURA17, PARK8, RIPK7, ROCO2 Tb-ERM (pLRRKtide) Fluorescein-ERM (LRRKtide) Inquire Inquire  
LTK TYK1 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
LYNA   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
LYNB   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
MAP3K1 0 MLK2, MST Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1      
MAP3K1 1 MLK3, SPRK Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1 Inquire Inquire  
MAP3K2 MEKK2, MEKK2B Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1 Inquire Inquire  
MAP3K3 MEKK3, MAPKKK3 Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1 Inquire Inquire  
MAP3K5 ASK1, MAPKKK5 Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1 Inquire Inquire  
MAP3K7 / MAP3K7 IP1 TAK1/TAB1 Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1 Inquire Inquire  
MAP3K8 COT1, EST, ESTF Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1 Inquire Inquire  
MAP3K9 MLK1, PRKE1 Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1 Inquire Inquire  
MAP4K2 GCK, BL44 Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire  
MAP4K4 HGK, NIK Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire  
MAP4K5 KHS, GCKR Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire  
MAPK1 ERK2, ERK Tb-pAT F2 [pThr71] GFP-AT F2      
MAPK10 JNK3 Tb-pAT F2 [pThr71] GFP-AT F2      
MAPK11 p38 beta, P38b, P382, SAPK2 Tb-pAT F2 [pThr71] GFP-AT F2      
MAPK12 p38 gamma, ERK6, SAPK3 Tb-pAT F2 [pThr71] GFP-AT F2      
MAPK13 p38 delta, SAPK4 Tb-pAT F2 [pThr71] GFP-AT F2      
MAPK14 p38 alpha, p38, Mxi2, SAPK2A Tb-pAT F2 [pThr71] GFP-AT F2      
MAPK3 ERK1, P44ERK1, PRKM3 Tb-pAT F2 [pThr71] GFP-AT F2      
MAPK8 JNK1, JNK, SAPK1 Tb-pAT F2 [pThr71] GFP-AT F2      
MAPK9 JNK2 Tb-pAT F2 [pThr71] GFP-AT F2      
MAPKAP K2 MK2 Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire Inquire  
MAPKAP K3 3PK, MAPKAP3 Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire Inquire  
MARK4 KIAA1860 Tb-pSe r (pPKC) Fluorescein-PKC      
MATK Hyl, CTK Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
MERTK cMER. MER Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
MET cMet, HGFR, RCCP2 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
MET M1250T cMet, HGFR, RCCP2 Tb-PY- 20 Fluorescein-Poly GT      
MINK1 MINK, B55, ZC3 Tb-pSe r (pPKC) Fluorescein-PKC      
MST1R RON, CDw136 Tb-PY- 20 Fluorescein-Poly GT      
MST4   Tb-pSe r (pPKC) Fluorescein-PKC      
MUSK   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
NEK2 NLK1, HsPK21 Tb-pSe r (pPKC) Fluorescein-PKC      
NLK FLJ21033, DKFZp761G1211 Tb-pc- Jun [pSer73] GFP-c- Jun      
NTRK1 TRKA, MTK, TRK Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
NTRK2 TRKB Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
NTRK3 TRKC Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
PAK4   Tb-pCr osstide Fluorescein-Crosstide      
PAK7 PAK5 Tb-pCr osstide Fluorescein-Crosstide      
PASK   Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire    
PDGFRA PDGFR alpha, CD140A, PDGFR2 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
PDGFRA D842V PDGFR alpha, CD140A, PDGFR2 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
PDGFRA T674I PDGFR alpha, CD140A, PDGFR2 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
PDGFRA V561D PDGFR alpha, CD140A, PDGFR2 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
PDGFRB PDGFR beta, JTK12, CD140B, PDGFR1 Tb-PY- 20 Fluorescein-Poly GT      
PHKG1   Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire  
PIM1   Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire Inquire  
PIM2   Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire Inquire  
PKN1 PRK1 Tb-pCR EB [pSer133] Fluore cein-CREBtide Inquire Inquire  
PRKACA PKA Tb-pCr osstide Fluorescein-Crosstide      
PRKCA PKC alpha Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire  
PRKCB1 PKC beta I Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire  
PRKCB2 PKC beta II Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire  
PRKCD PKC delta Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire  
PRKCE PKC epsilon Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire  
PRKCG PKC gamma Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire  
PRKCH PKC eta Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire  
PRKCI PKC iota Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire  
PRKCN PKD3, PRKG3, EPK2, PKC nu Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire Inquire  
PRKCQ PKC theta Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire  
PRKCZ PKC zeta Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire  
PRKD1 PKD, PKC mu Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire Inquire  
PRKD2 PKD2, HSPC187 Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire Inquire  
PRKG1 PGK, CGK1, FLJ36117 Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire Inquire  
PRKG2 PKG2, PRKGR2, cGKII Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire Inquire  
PRKX PKX1 Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire  
PTK2 FAK, FADK, FAK1 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
PTK2B FAK2, CAKB, PYK2 Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
PTK6 Brk Tb-PY- 20 Fluorescein-Poly GT      
RAF1 (cRaf) Y340D Y341D   Tb-pMA P2K1 [pSer217/221] Fluorescein-MAP2K1      
RET   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
RET V804L   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
RET Y791F   Tb-PY- 20 Fluorescein-Poly GT      
ROCK1   Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire  
ROCK2 KIAA0619 Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire  
ROS1 ROS, MCF3 Tb-PY- 20 Fluorescein-Poly GT      
RPS6KA 1 RSK1, HU-1 Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire  
RPS6KA 2 RSK3, HU-2 Tb-pSe r (pPKC) Fluorescein-PKC Inquire Inquire  
RPS6KA 3 RSK2, HU-3 Tb-pCr osstide Fluorescein-Crosstide Inquire Inquire  
RPS6KA 4 MSK2, RSK-B Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire Inquire  
RPS6KA 5 MSK1, RLPK Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire Inquire  
RPS6KB 1 p70S6K, STK14A, S6K Tb-pCr osstide Fluorescein-Crosstide      
SGK SGK1 Tb-pSe r (pPKC) Fluorescein-PKC      
SGK2   Tb-pSe r (pPKC) Fluorescein-PKC      
SGKL SGK3 Tb-pSe r (pPKC) Fluorescein-PKC      
SRC ASV Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
SRCN1   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
SRMS SRM Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
STK22D TSSK1, FKSG81, SPOGA4 Tb-pCR EB [pSer133] Fluorescein-CREBtide Inquire Inquire  
STK24 MST3, STK3 Tb-pSe r (pPKC) Fluorescein-PKC      
STK25 YSK1, SOK1 Tb-pSe r (pPKC) Fluorescein-PKC      
SYK   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
TAOK2 TAO1, PSK, TAO2, KIAA0881 Tb-pSe r (pPKC) Fluorescein-PKC      
TAOK3 JIK Tb-pCr osstide Fluorescein-Crosstide      
TBK1 NAK, T2K, FLJ11330 Tb-IkBa
Fluorescein-IKK
TEC PSCTK4 Tb-PY- 20 Fluorescein-Poly GT      
TEK Tie2, VMCM, CD202B Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
TGFBR1 ALK5 Tb-PY- 20 Fluorescein-Poly GT Inquire Inquire  
TNK2 ACK Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
TTK MPS1L1 Tb-PY- 20 Fluorescein-Poly GT      
TXK   Tb-PY- 20 Fluorescein-Poly GT      
TYK2 JTK1 Tb-pST AT1 [pTyr701] GFP-ST AT1      
TYRO3 DKT, BRT, TIF Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
YES1 Yes, C-YES, P61-Yes Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT  
ZAP70   Tb-PY- 20 Fluorescein-Poly GT Eu-PY- 20 Alexa Fluor 647-Poly GT