CHO Cell Culture Media
Simplify downstream protein purification with serum-free CHO Medium
- Get optimal expression of recombinant proteins
- Select from chemically defined, protein-free, and serum-free formulations
- Choose media for suspension cells
| Low-protein (<100 mg/L), serum-free alternative for CHO cell line growth and production | Optimizes growth of parental DHFR– CHO cells | Consistent production medium | High-performing, leaner medium designed for fed-batch optimization | Selectively fortified for fastest fed-batch process development starting point | |
|---|---|---|---|---|---|
| CHO cell line | Various CHO dependent on experience | DG44, DXB11, or other CHO | Various CHO dependent on experience | DG44, DXB11, or other CHO | K1, CHO-S®, GS |
| Culture type** | Suspension | Suspension | Suspension | Suspension | Suspension |
| Classification | Serum-Free, contains proteins of animal origin | Serum-Free, Protein-Free, Animal Origin-Free, Chemically Defined* | Serum-Free, Protein-Free, Animal Origin-Free, Chemically Defined* | Serum-Free, Protein-Free, Animal Origin-Free, Chemically Defined* | Serum-Free, Protein-Free, Animal Origin-Free, Chemically Defined* |
| Relative titer in batch culture*** | Medium | N/A (no titer since cells are parental) | High | Higher | Highest |
| Culture duration | Average <14 days | Average <14 days | ≥14 days with feeding | ≥14 days with feeding | ≥20 days with feeding |
| Recommended Feed | CHO CD EfficientFeed™ Kit | CHO CD EfficientFeed™ Kit | CHO CD EfficientFeed™ B | CD EfficientFeed™ A | CD EfficientFeed™ C |
| DMF reference requests | Contact Technical Services | Contact Technical Services | Contact Technical Services | Contact Technical Services | Contact Technical Services |
| List price | 1L=$87.00 | 1L=$87.00 | 1L=$84.00 | 1L=$76.00 | 1L=$93.00 |
| Form | Liquid | Liquid | Liquid & AGT™ | Liquid & AGT™ | Liquid |
| Liquid sizes | 500mL 1L |
1L | 500ml 1L 10L 20L |
1L | 1L |
| AGT™ sizes | N/A | N/A | 1L (24.26g) 10L (242.60g) 100L (2426.0g) 10kg |
1L (19.318g) 10L (193.180g) 100L (1931.800g) 10kg |
N/A |
| Option to customize | Yes | Yes | Yes | Yes | Yes |
* Chemically Defined means that the product does not contain any proteins or undefined components like hydrolysates or hydrolysate fractions.
** Products are intended for preparation for and use in stable cell line protein expression. See FreeStyle™ expression products for transient transfection.
*** Dependent on cell line clone