CHO Supplement
EfficientFeed™ supplements have been designed to improve CHO (Chinese hamster ovary) fed-batch culture processes for the production of recombinant proteins in suspension culture. All of these products are chemically defined and protein-free. They contain no components of animal origin.
| Best place to start if using CD OptiCHO™ base medium | Best place to start if using CD CHO base medium | Most concentrated feed with GS supplement "built in" |
Fastest way to find feeding conditions that maximize CHO protein expression | |
|---|---|---|---|---|
|
Complete Feed -
EfficientFeed™ A |
Complete Feed - EfficientFeed™ B |
Complete Feed - EfficientFeed™ C |
Complete Feed - EfficientFeed™ Kit |
|
| Matching Gibco® Medium | CD OptiCHO™ medium | CD CHO medium | CD FortiCHO™ medium | CD OptiCHO™ or CD CHO |
Select if non-Gibco® medium has |
![]() |
![]() |
![]() |
|
Select if non-Gibco® medium has |
![]() |
|||
| Classification | Serum-Free, Protein-Free, Animal Origin-Free, Chemically Defined* | Serum-Free, Protein-Free, Animal Origin-Free, Chemically Defined* | Serum-Free, Protein-Free, Animal Origin-Free, Chemically Defined* | Serum-Free, Protein-Free, Animal Origin-Free, Chemically Defined* |
| Titer in fed-batch vs. batch | 2–5 fold increase | 2–5 fold increase | 2.5–6 fold increase | 2–5 fold increase |
| Culture duration | >14 days in fed-batch |
>14 days in fed-batch |
>14 days in fed-batch |
>14 days in fed-batch |
| Volume of addition | 10–50% of working volume | 10–50% of working volume | 10–40% of working volume | 10–50% of working volume |
| List price | 1 L = $138 | 1 L = $138 | 1 L = $147 | Kit = $268 |
| Approximate list price per liter of culture | $55.00 | $55.00 | $44.00 | $55.00 |
| GS system supplement | Add GS supplement | Add GS supplement | GS supplement not needed, already built in |
Add GS supplement |
| DMF reference requests |
Contact Technical Services | Contact Technical Services | Contact Technical Services | Contact Technical Services |
| Form | Liquid & AGT™ | Liquid & AGT™ | AGT™ | Liquid |
| Liquid sizes | 1 L | 1 L | N/A | 1 L |
| AGT™ sizes | 10 L (651 g) 100 L (6,510 g) |
10 L (541.5 g) 100 L (5,415 g) |
1 L (79.6 g) 10 L (796 g) 100 L (7,960 g) |
N/A |
| Option to customize | Yes | Yes | Yes | Yes |
* Chemically Defined means that the product does not contain any proteins or undefined components like hydrolysates or hydrolysate fractions