siRNA Transfection
![]() |
Gene silencing or knockdown can be done easily now in most cell types, but there are a few things you need to consider before starting:
|
Which siRNA transfection product is right for you?
| Order now | Order now | Order now | Order now | |
| High efficiency versatile reagent for a wide range of common cell types | Most efficient versatile reagent for the widest range of cell types including difficult-to-transfect cells | Most efficient reagent for siRNA/miRNA delivery. Efficient gene knock-down. | High-efficiency electroporation for all cell lines. Best for Primary & Stem Cells | |
|---|---|---|---|---|
| Lipofectamine™ 2000 Learn more |
Lipofectamine™ 3000 Learn more |
Lipofectamine™ RNAiMAX Learn more |
Neon™ Transfection System Learn more |
|
| Top Selling Format | Catalog #11668-019 - 1.5 mL | Catalog #L3000-015 - 1.5 mL | Catalog #13778-075 - 0.75 mL | Catalog #MPK5000S |
| Sample Type |
|
|
|
|
| Transfection Efficiency | High | Superior | Superior | Maximal |
| Cell Viability | High | Superior | Superior | Good |
| Protocol | Download Protocol | Download Protocol | Download Protocol | Download Protocol |