Competent Cells for Routine Cloning/Subcloning
Chemically competent and high transformation efficiency cells that do not require an electroporator.
Which competent cell strain is right for you?
| Economical; for routine subcloning | Mid-range transformation efficiency | High-efficiency, best all around | Highest efficiency, ideal for low starting DNA | High efficiency, fast growing | |
|---|---|---|---|---|---|
|
|
|
|
|
|
|
| Top Seller | ![]() |
![]() |
|||
| Transformation Efficiency of pUC19 | >1x106 cfu/ug | >1x108 cfu/ug | >1x109 cfu/ug | >5x109 cfu/ug (highest) |
>1x109 cfu/ug |
| One Shot format | ![]() |
![]() |
![]() |
||
| T1 & T5 Phage resistant (ton A) | ![]() |
![]() |
|||
| Time to pick colonies | Overnight | Overnight | Overnight | Overnight | Same day |
| Time to purify plasmid DNA after inoculation | Overnight | Overnight | Overnight | Overnight | As soon as 4 hrs |
| Blue/White Screening | ![]() |
![]() |
![]() |
![]() |
![]() |
| Efficiently transform unmethylated DNA (hsd R) | ![]() |
![]() |
|||
| Efficiently transform methylated DNA (mcr A) | ![]() |
![]() |
|||
| Generate ssDNA (Contains F' Episome) | ![]() |
||||
| Kit contains | 4 tubes x 500 µL | 5 tubes x 200 µL | 20 tubes x 50 µL | 20 tubes x 50 µL | 20 tubes x 50 µL |
| Order | Order | Order | Order | Order |