Obtaining high-quality, intact DNA is often the first and most critical step in many fundamental molecular biology applications, such as DNA cloning, sequencing, PCR, and electrophoresis. Isolating intact total DNA from tissue samples varies in difficulty with the physical and biochemical nature of the tissue. For example, many DNA extraction methods will work equally well for tissues such as liver, but fibrous tissues such as heart, fatty tissues such as brain, and nuclease-rich tissues like spleen present challenges for DNA isolation. We offer a wide range of DNA extraction from tissue kits, plus the support you may need to get high yields of pure, intact DNA from almost any type of tissue.
DNA Extraction From Tissue
Which Genomic DNA Extraction From Tissue Kit is right for you?
| Order now | Order now | Order now | Order now |
|
| Process the largest amount of tissue | Fast isolation of gDNA from a variety of samples | High -yield, high-purity gDNA in a plate format | Rapid and automated extraction of DNA | |
|---|---|---|---|---|
| DNAzol™ Reagent | PureLink™ Genomic DNA Mini Kit | PureLink™ Pro 96 Genomic DNA Purification Kit | MagMAX™-96 DNA Multi-Sample Kit | |
| Tissue starting material | Up to 50 mg | Up to 25 mg | Up to 25 mg | Up to 25-50 mg |
| Yield | Up to 250 µg | 5-10 µg | 5-10 µg | 10-80 µg |
| Isolation method | Organic extraction | Silica membrane | Filter plate | Magnetic beads |
| High-throughput compatible | No | No | Yes | Yes |
| Compatible applications | Cloning, qPCR, sequencing | Cloning, qPCR, sequencing, genotyping | Cloning, qPCR, sequencing, genotyping | Cloning, qPCR, sequencing, genotyping |
| Prep time | 10-30 min | 15 min | 35 min | 45 min |
| Prep size | 100 mL | 50, 250 preps | 4 x 96 preps | 50, 96, 500, 2500 preps |
Resources
Nucleic Acid Purification and Analysis Support Center
Find tips, troubleshooting help, and resources for your nucleic acid purification & analysis applications.
DNA Purification Brochure
RNA Isolation & Purification Brochure
- Protocol Videos—Videos to help you isolate nucleic acids using a variety of techniques.
- DNA & RNA Selection Guide—Find the right purification products.
- Webinars—View our library of recorded nucleic acid isolation webinars.