DNA Gels for Next-Generation Sequencing
Achieving reliable sequencing data while preserving the integrity of the original DNA sample is important for constructing your DNA library. We've developed a wide selection of separation technologies for preparing DNA fragment libraries for next-generation sequencing—ranging from UltraPure™ agarose to the E-Gel® SizeSelect™ system, which has been successfully used in two sequencing platforms.
Which DNA gel is right for you?
| Standard solution for NGS | Improved solution for NGS | Ultimate solution for NGS | |
|---|---|---|---|
| Order Now | Order Now | Order Now | |
| UltraPure™ Agarose-1000 Learn more › |
E-Gel® NGS Learn more › |
E-Gel® SizeSelect™ Learn more › |
|
| Top seller | ✓ | ||
| Prep time | 30 min | Ready to use | Ready to use |
| Run time | 60 min | 30 min | 10 min |
| Purification time | 30 min | 30 min | Ready to use |
| Ease of obtaining sequence template* | Moderate effort (band excision & purification) | Moderate effort (band excision & purification) | Easy (direct from well) |
| # of lanes | Variable | 11 | 9 |
| % gels available | Variable | 0.8% | 2% |
| Gel dye options | Add your own dye | SYBR® Safe | Proprietary Stain |
| Pack sizes available | 100 g | 10 gels | 10 gels |
* Avoid manual DNA band excision to increase precision and accuracy of NGS applications with results that are easily reproducible across experiments. Learn more ›