The mirVana™ miRNA Isolation Kits use a rapid procedure to isolate total RNA, including small RNAs such as microRNA (miRNA), small interfering RNA (siRNA), and small nuclear RNA (snRNA), from tissues and cells. The mirVana™ miRNA Isolation Kit employs organic total RNA extraction followed by purification on a GFF (glass fiber filter) using specialized binding and wash solutions.
The kit effectively recovers all RNA:
- Large mRNA and ribosomal RNA down to 10-mers
- Virtually all cell and tissue types