QuantStudio 3 Real-Time PCR System
Intuitive and easy to use for all levels of experience
Now you can get up and running quickly with the Applied Biosystems QuantStudio 3 Real-Time PCR System, a simple and affordable real-time PCR solution designed for users with all levels of experience. Get connected to your data easier than before with a real-time PCR systems that offers an interactive touch screen and the option to choose Web browser-based software enabled through the Thermo Fisher Cloud.
The QuantStudio 3 Real-Time PCR System is available with 96-well thermal blocks with either 0.1 mL or 0.2 mL well volume.
- Download brochure
- Contact us
- Telephone: 800 327 3002, option 4, then option 3 (toll-free in the US only)
- Telephone 00 800 5345 5345, option 3 (toll-free in Europe, Middle East and South Africa only)
Get started quickly—The factory-calibrated system is delivered in a user-friendly package, enabling quick installation and immediate use.
Skip the learning curve—With preoptimized templates and an intuitive interface that requires minimal training, the QuantStudio Design and Analysis Software is available in desktop and Web browser-based format.
Helps save time with efficient experiment runs—Applied Biosystems VeriFlex technology offers 3 independent temperature zones for PCR optimization, and fast thermal cycling enables you to get results in as little as 30 minutes.
Results you can trust—Applied Biosystems OptiFlex technology with white LED and 4 coupled channels enables optimal well-to-well and instrument-to-instrument accuracy.
Get started with a limited budget—Get the modern industrial design and superior functionality you have come to know with the Applied Biosystems QuantStudio family of high-performance instruments. The QuantStudio 3 instrument is the perfect entry-level system and allows you to stretch your budget.
Get connected to your data more easily and quickly than ever before, with a real-time PCR system that offers an interactive touch screen for a modern usability experience similar to mobile devices and the option to choose Web browser–based software enabled through the Thermo Fisher Cloud. With the cloud, you can:
- Remotely monitor your runs and access data anytime, anywhere*
- Analyze sophisticated data sets quickly
- Store results in a secure data storage space
- Share and collaborate with colleagues at your institution or around the world
For peace of mind, the Thermo Fisher Cloud offers high industry-standard security, so your work remains secure. We have partnered with Amazon Web Services® (AWS) to offer a robust platform.
*Where the Web is accessible
| Repair Center service plans | On-site service plans | |||
|---|---|---|---|---|
| Service plans at a glance | AB Repair Center Support Plus | AB Maintenance | AB Assurance | AB Complete |
| On-site response time | Target 2 business days** | 2 business days* | Next business day* | |
| Scheduled on-site planned maintenance (PM) | Yes | Yes | Yes | |
| Remote diagnostics | Yes | Yes | Yes | Yes |
| Parts, labor, and travel for repair included | Yes | Yes | Yes | |
| Computer repair and replacement included | Yes | Yes | Yes | |
| Priority access to Tech Support Mon–Fri, 8 a.m.—5 p.m. local time |
Yes | Yes | Yes | |
| Priority access to Remote Service Engineer | Yes | Yes | Yes | |
| Loaner instrument issued during repair (Repair Center plans only) | Yes | Yes | ||
*Availability limited in some geographic ares.
**After receipt of a purchase order.
QuantStudio Family of Real-Time PCR and Digital PCR Systems
Product brochure: QuantStudio 3 and QuantStudio 5 Real-Time PCR Systems
Your Innovative Research: QuantStudio 5 Real-Time PCR System
Application Note: QuantStudio 3 and 5 Real-Time PCR Systems
Your Innovative Research: Expression of oncogenic microRNAs
Application Note: Protein Thermal Shift Technology
Get started quickly—The factory-calibrated system is delivered in a user-friendly package, enabling quick installation and immediate use.
Skip the learning curve—With preoptimized templates and an intuitive interface that requires minimal training, the QuantStudio Design and Analysis Software is available in desktop and Web browser-based format.
Helps save time with efficient experiment runs—Applied Biosystems VeriFlex technology offers 3 independent temperature zones for PCR optimization, and fast thermal cycling enables you to get results in as little as 30 minutes.
Results you can trust—Applied Biosystems OptiFlex technology with white LED and 4 coupled channels enables optimal well-to-well and instrument-to-instrument accuracy.
Get started with a limited budget—Get the modern industrial design and superior functionality you have come to know with the Applied Biosystems QuantStudio family of high-performance instruments. The QuantStudio 3 instrument is the perfect entry-level system and allows you to stretch your budget.
Get connected to your data more easily and quickly than ever before, with a real-time PCR system that offers an interactive touch screen for a modern usability experience similar to mobile devices and the option to choose Web browser–based software enabled through the Thermo Fisher Cloud. With the cloud, you can:
- Remotely monitor your runs and access data anytime, anywhere*
- Analyze sophisticated data sets quickly
- Store results in a secure data storage space
- Share and collaborate with colleagues at your institution or around the world
For peace of mind, the Thermo Fisher Cloud offers high industry-standard security, so your work remains secure. We have partnered with Amazon Web Services® (AWS) to offer a robust platform.
*Where the Web is accessible
| Repair Center service plans | On-site service plans | |||
|---|---|---|---|---|
| Service plans at a glance | AB Repair Center Support Plus | AB Maintenance | AB Assurance | AB Complete |
| On-site response time | Target 2 business days** | 2 business days* | Next business day* | |
| Scheduled on-site planned maintenance (PM) | Yes | Yes | Yes | |
| Remote diagnostics | Yes | Yes | Yes | Yes |
| Parts, labor, and travel for repair included | Yes | Yes | Yes | |
| Computer repair and replacement included | Yes | Yes | Yes | |
| Priority access to Tech Support Mon–Fri, 8 a.m.—5 p.m. local time |
Yes | Yes | Yes | |
| Priority access to Remote Service Engineer | Yes | Yes | Yes | |
| Loaner instrument issued during repair (Repair Center plans only) | Yes | Yes | ||
*Availability limited in some geographic ares.
**After receipt of a purchase order.
QuantStudio Family of Real-Time PCR and Digital PCR Systems
Product brochure: QuantStudio 3 and QuantStudio 5 Real-Time PCR Systems
Your Innovative Research: QuantStudio 5 Real-Time PCR System
Application Note: QuantStudio 3 and 5 Real-Time PCR Systems
Your Innovative Research: Expression of oncogenic microRNAs
Application Note: Protein Thermal Shift Technology