Transfer Proteins for Western Blot
One of the key steps in the western blot workflow is the transfer of proteins from the polyacrylamide gel after electrophoresis to the nitrocellulose or polyvinylidene difluoride (PVDF) membrane so that specific proteins can be detected using immunodetection techniques.
Methods for achieving this protein transfer include capillary transfer or electrophoretic transfer. We have developed electrophoretic transfer systems using wet, semi-dry, and dry methods outlined below. Use the table below to select the appropriate transfer method for your western blotting needs.
Which western transfer system is right for you?
| Traditional wet transfer | Semi-dry transfer | Dry transfer | |||
|---|---|---|---|---|---|
![]() Mini Blot Module |
Pierce Power Blotter |
iBlot 2 Gel Transfer Device |
|||
| Transfer time | 60–120 min | 60 min | 30–60 min | 5–10 min | 7 min |
| Capacity of device | 1–2 mini gels | 1 mini gel (per module) or 2 mini gels (two modules per tank) | 4 mini-gels or 2 midi-gels |
4 mini gels or 2 midi gels |
2 mini gels or 1 midi gel |
| Blotting area | 9 x 9 cm | 9 x 9 cm | 21 x 21 cm | 21 x 22.5 cm | 8.5 x 13.5 cm |
| Requires transfer buffer | Yes (1,000 mL) | Yes, (200–400 mL per module) | Yes (500 mL) | Yes (50 mL per mini gel or 100 mL per midi gel) | No |
| Power supply | External | External | External | Internal | Internal |
| Order Now | Order Now | Order Now | Order Now | Order Now | |
Resources |
NEW | Protein Electrophoresis and Western Blotting Support Center Find tips, troubleshooting help, and resources for your western workflow. |
|
Pierce Power Blotter
iBlot 2 Gel Transfer Device