Perform analysis of total halogens and sulfur in difficult-to-analyze samples
Explore even more sample types and spark new insights with the Thermo Scientific Cindion Combustion Ion Chromatography (C-IC) System. This fully-integrated C-IC system can be used to screen for potentially corrosive and toxic halogens and sulfur, helping you ensure high-quality standards to safeguard products and the environment. Whether you are analyzing solid, liquid, or gas samples, experience automated workflows with robust performance and dependable reliability from a single-source system.
Highlights of the Cindion C-IC System
Multiple system configurations
Measure halogens and sulfur in solid, liquid, and gas samples. The Cindion C-IC System is available in various configurations to meet your requirements.
Single-source solution
Experience a seamless, end-to-end C-IC workflow. Whether you are analyzing solid, liquid, or gas samples, we offer all the necessary consumables, software, and support to fully outfit your C-IC system.
Sensitive PFAS screening
With a sample flow path that minimizes use of materials that can contribute to detectable PFAS levels, the Cindion C-IC System provides highly sensitive detection to measure adsorbable organic fluorine (AOF) as a PFAS screening method.
Single-software control
Control and monitor your combustion IC analysis with a single software. From sample combustion to IC analysis, Thermo Scientific Chromeleon Chromatography Data System (CDS) brings out the full power of the Cindion C-IC System.
Industry-leading IC system
The Cindion C-IC System takes advantage of industry-leading IC technology. Features such as electrolytic suppression and Reagent-Free IC (RFIC) eluent generation enhance consistency and simplify IC analysis.
Experience and support
We offer an extensive library of C-IC application notes for multiple sample types in the industrial, environmental, and food & beverage industries.
Our general purpose product lines, including LC-MS products, are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks. They are designated for Research Use Only Applications.
For Research Use Only - Not For Use In Diagnostic Procedures.