Search
| Organism Type | Insect | Derived from Drosophila (fruit fly) embryo; doubling time is 20 to 24 hours. These cells are also known as S2 or SL2 cells. These cells can be grown in serum-supplemented or serum-free media in both adherent or suspension conditions. ATCC CRL-1963 |
| Tissues | Embryo | |
| Phenotype | Either | |
| Primary | no | |
| Application | Cell Culture / Growth Conditions,Nucleic Acid Purification,Protein Expression,RNAi,Transient Transfection |