Example of recommended starting conflucence of PSCs (Day 1)
We recommend starting with 15-30% confluent PSCs for induction. Below are examples showing optimum confluency before induction (24 h after seeding). Induction at higher starting confluency will lead to higher number of floating cells.