Search
Find valuable information.
Optimize your experiments to get the best results. We’ve compiled a detailed knowledgebase of the top tips and tricks to meet your research needs.
View the relevant questions below:
Having problems with your experiment? Visit our
Clones should be streaked out on the appropriate selective media to single colonies.
Sorry, we do not offer the empty pCMVSPORT6 vector.
When an order is placed, our in-house manufacturing group will try to grow up the clone. If the clone does not grow, they will make a second attempt to grow the clone, after which they will inform the customer that it is a “no grow”.
We recommend choosing the Ultimate™ ORF clones if quality and sequence are your main priority. Further, these clones are provided in a Gateway® vector backbone, so it is easy to shuttle the insert into multiple host systems. The Full-length human cDNA clones and MGC clones are only 5’ and 3’ end-sequenced, but Full-length human cDNA clones offer the advantage of being in a Gateway® entry vector backbone. MGC clones are available for human, mouse, and rat species; Ultimate™ ORF clones are available for human and mouse species and Full-Length human cDNA clones are available only for human species.
The backbone vector that is used for the Full-length human cDNA clone collection is pCMV-SPORT6. It is a Gateway-adapted mammalian expression vector where the insert is driven by the CMV promoter and flanked by attB1 and attB2 sites. The construct can be directly used for expression of the gene in mammalian cells.
Please go to the NIH Mammalian Gene Collection Consortium website at http://mgc.nci.nih.gov
The cDNA clones we offer are Ultimate™ ORF clones, Full-Length human cDNA clones, MGC clones, and GeneStorm® Expression-Ready clones. The Ultimate™ ORF Clones, Full-length human cDNA clones, and the GeneStorm® Expression-Ready clones are made in-house, whereas the MGC clones are derived from external sources and distributed by Life Technologies. The Ultimate™ ORF clones are fully-sequenced and guaranteed to match GenBank sequence information 100% at the amino acid level, whereas the sequence of the other cDNA clones we offer is not guaranteed.
The backbone vector that is used is pENTR221, which is a Gateway® Entry vector. The insert (an open reading frame that begins with ATG and ends with a TAG stop codon) is flanked by attL1 and attL2 sites. The construct is ready for Gateway® LR recombination with a DEST vector containing attR1 and attR2 sites. It is compatible with N-terminal tagged DEST vectors but not with C-terminal tagged DEST vectors. To express the C-terminal tag, the Tag-On-Demand technology would have to be used or the TAG stop codon would have to be mutated. In place of the Tag-On-Demand adenovirus product we sold previously, we offer the vector expressing the tRNA suppressor as a custom service. If you are interested in this service, please e-mail custom.services@lifetech.com .
GeneStorm® Expression-Ready human clones are made using the highly-efficient ACE™ (Accelerated Cloning and Expression) process. This process applies PCR cloning, DNA sequencing, and protein expression testing in a high-throughput format. Human genes are amplified using gene-specific primer sets designed to amplify only full-length open reading frames. The PCR products are then TOPO-cloned into the GeneStorm® (GS) mammalian expression vectors, pcDNA3.1/GS.
BAC clones are genomic clones containing very large inserts (>100 kb) that may contain several genes, whereas cDNA clones only contain a single open reading frame of a gene. BAC clones are used for promoter and intron/exon analysis, chromosome localization, and making transgenic mice.
We recommend using the PureLink® HiPure plasmid DNA purification kits in mini, midi or maxi formats (Cat. Nos. K210002, K210003, K210004, K210005, K210006, K21007). There is a modified protocol in the appendix of the manual that will assist in increasing yield when purifying extremely large plasmids. We also offer a PureLink® HiPure BAC Buffer Kit (Cat. No. K210018) to supply the additional buffers required for the BAC purification protocol.
BAC/PAC clones cannot be found in the CloneRanger™ tool by gene name or Genbank accession number, and Life Technologies does not have the sequence or insert identity for these clones.
To find/order an individual BAC/PAC clone, you need the Clone Address (name of the library and the Clone ID or location of the clone on the plate). For example: CTD-2320O4 = CITB D library, Clone ID 2320O4, where 2320 is the Plate #, O is the Row, and 4 is the Column #. To order this clone, you have to provide the Cat # (96012) and Clone address (CTD-2320O4).
For Research Use Only. Not for use in diagnostic procedures.