In a time when getting answers quickly is critical, Thermo Fisher Scientific is leveraging its scale and resources to help the world move forward. With innovative solutions and a reliable supply chain already helping meet the unprecedented demand for trusted COVID-19 testing, we continue to expand our high-quality solutions and accelerate innovation with urgency, addressing current and future challenges.
Explore this section to find out how we’ve contributed to the fight against COVID-19 through the use of our chemicals across various applications, from qPCR testing to hand sanitizers.
Sample Collection via Antigen Rapid / Lateral Flow Test Kits
Product Name
Application
Product Example
Guanidine Thiocyanate
  • Solubilize proteins by increasing solubility of hydrophobic peptides and proteins
  • Denaturing proteins by disrupting hydrogen bonds which hold the protein in its unique structure (Lysis Buffer).
Guanidine Thiocyanate, 99.5%
Polyethylene glycol dodecyl ether
  • Detergent to solubilize protein
Polyethylene glycol dodecyl ether, >95% (Thesit®)
Proteinase K
  • Enzyme used to cleave peptide bond between amino acids
  • Effectively inactivates DNAse & RNAse
  • Commonly used for nucleic acid preparation workflow
Proteinase K, for molecular biology
Skim Milk
  • Component in blocking buffers to prevent non-specific binding of proteins-of-interest
  • Skim Milk Powder
    Tricine
  • Used for resuspension of cell pellets
  • Tricine, 99%
    Tris
  • Organic compound that is extensively used in buffer solutions (TAE and TBE buffers)
  • Commonly used as Elution Buffer
  • Tris(hydroxymethyl)aminomethane, 99.9%
    qPCR Testing: Sample Preparation and Analysis
    Already using an existing Thermo Fisher Scientific product of instrument? For example, MagMAX kits, Kingfisher, taqpath, etc., we have complementing solvents and solutions such as the 80% Molecular Biology grade Ethanol for you. The table below outlines our top selling solvents in the biopharmaceutical and biotechnology industries.
    Water for Life Sciences Applications
    Fisher Bioreagents waters are ideal for many fundamental procedures such as PCR, electrophoresis, DNA sequencing, and as buffers for enzymatic analyses. Their low metal content ensures minimal free ions, which lets you prepare optimized enzymatic reaction buffers by adjusting the concentration of the appropriate metal ion cofactor(s).
    DescriptionSizeCatalog NumberDEPC-TreatedAutoclaved0.2 µm Filtered0.03µm FilteredDeionized
    Water, DNA Grade
    1 L
    BP2470-1 
    Water, Nuclease Free
    50 mL 100 mL
    BP2484-50
    BP2484-100
    Water, Biotech Grade
    4 L 20 L
    BP2485-4
    BP2485-20
    Water, Molecular Biology Grade
    100 mL 1 L 4 L 10 L 20 L
    BP2819-100
    BP2819-1
    BP2819-4
    BP2819-10
    BP2819-20
    Water, Microbial Cell Culture Grade
    100 mL 500 mL 1 L
    BP2820-100
    BP2820-500
    BP2820-1
    Water, qPCR Grade
    1 L 10 L
    BP2825-1
    BP2825-10
    Water, RNA Grade
    1 L
    BP561-1
    Water for Life Sciences Applications – Activity Specifications
    Description
    Size
    Catalog Number
    Total Aerobic Microbial Count
    Total Yeast and Mold Count
    Endotoxin
    DNase Free
    RNase Free
    Proteaase Free
    Nickase Free
    Human Genomic DNA
    E. coli Genomic DNA
    Water, DNA Grade
    1 L
    BP2470-1     
    Water, Nuclease Free
    50 mL 100 mL
    BP2484-50
    BP2484-100
    Water, Biotech Grade
    4 L 20 L
    BP2485-4
    BP2485-20
    Water, Molecular Biology Grade
    100 mL 1 L 4 L 10 L 20 L
    BP2819-100
    BP2819-1
    BP2819-4
    BP2819-10
    BP2819-20
    Water, Microbial Cell Culture Grade
    100 mL 500 mL 1 L
    BP2820-100
    BP2820-500
    BP2820-1
    Water, qPCR Grade
    1 L 10 L
    BP2825-1
    BP2825-10
    Water, RNA Grade
    1 L
    BP561-1     
    Ethanol for Nucleic Acid and Protein Purification
    Molecular biology grade ethanol options now include 100% ethanol and 70%, 80%, and 96% solutions.
    Ethanol products are tested to verify the absence of DNase, RNase, protease, and other hydrolytic enzymes that can degrade target biomolecules during isolation and purification. These solutions also work well in combination with commercial mini-prep kits or traditional extraction methods.
    Test70% Ethanol80% Ethanol96% EthanolAbsolute Ethanol(100%)
    BrandFisher BioreagentsThermo ScientificFisher Bioreagents Fisher Bioreagents
    Cat. No.BP8201T08204K7BP8202BP2818
    AppearanceClear, ColorlessClear, ColorlessClear, Colorless Clear, Colorless
    Color(APHA)<10<10<10 <10
    Residue After Evaporation<0.001%BP2820-100
    BP2820-<0.001%
    BP2820-1
    <0.001%<0.001%
    Substances Darkened by Sulfuric AcidPass TestPass TestPass TestPass Test
    Substances Reducing PermanganatePass TestPass TestPass Test Pass Test
    Assay(GC-TCD)
    Ethanol
    Water
    Methanol
    IPA
    68%-72%
    28%-32%
    NA
    NA
    78%-82%
    18%-22%
    NA
    NA
    94%-98%
    2%-6%
    NA
    NA
    ≥99.5%
    ≤0.2%
    <0.1% NA
    0.003%
    DNaseNot DetectedNot DetectedNot DetectedNot Detected
    RNaseNot DetectedNot DetectedNot DetectedNot Detected
    ProteaseNot DetectedNot DetectedNot DetectedNot Detected
    Nucleic Acid Extraction: Our response in COVID-19 related activities
    Find out more ››
    Guanidine Thiocyanate: Lysis buffers for nucleic acid extraction
    Find out more ››