Search
Catalog Number(s)
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Inorganic Salts | |||
| Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 1400.0 | 12.612613 |
| Magnesium Chloride (MgCl2-6H2O) | 203.0 | 1000.0 | 4.9261084 |
| Magnesium Sulfate (MgSO4-7H2O) | 246.0 | 1000.0 | 4.0650406 |
| Potassium Chloride (KCl) | 75.0 | 4000.0 | 53.333332 |
| Potassium Phosphate monobasic (KH2PO4) | 136.0 | 600.0 | 4.4117646 |
| Sodium Chloride (NaCl) | 58.0 | 80000.0 | 1379.3103 |
| Sodium Phosphate dibasic (Na2HPO4-7H2O) | 268.0 | 900.0 | 3.358209 |
| Other Components | |||
| D-Glucose (Dextrose) | 180.0 | 10000.0 | 55.555557 |
| Phenol Red | 398.0 | 100.0 | 0.2512563 |
1. Hanks, J.H. and Wallace, R.E. (1949) Relation of Oxygen and Temperature in the Preservation of Tissues by Refrigeration. Proc. Soc. Exp. Biol. Med., 71:196.
24020 - HBSS, calcium, magnesium
Cat No:
24020083
,
24020091
,
24020117
,
24020133
,
24020141
14025 - HBSS, calcium, magnesium, no phenol red
Cat No:
14025050
,
14025076
,
14025092
,
14025100
,
14025126
,
14025134
14065 - HBSS, 10x, calcium, magnesium, no phenol red
Cat No:
14065049
,
14065056
,
14065072