Live bovine pulmonary artery endothelial cells (BPAEC) labeled with LysoTracker® Red and Hoechst 33342.
Live bovine pulmonary artery endothelial cells (BPAEC) were labeled with LysoTracker® Red DND-99 (Cat. no. L7528), a BODIPY® derivative, and Hoechst 33342 (Cat. no. H1399, H3570, H21492). The cells were then imaged by fluorescence and differential interference contrast (DIC) microscopy.
Live bovine pulmonary artery endothelial cells (BPAEC) were labeled with LysoTracker® Red  DND-99 (Cat. no. L7528), a BODIPY® derivative, and Hoechst 33342 (Cat. no. H1399, H3570, H21492). The cells were then imaged by fluorescence and differential interference contrast (DIC) microscopy.