Search
Our software is built using specifications gathered from laboratories around the world. Along with HistoTrac Core Software, the additional modules enable the laboratory to maintain lab-specific records pertaining to purchasing, supply inventory, and mailings. Use these modules to assist in the organization of your work, to reduce the need for extra paper and spreadsheets, and to eliminate hand-written labels.
The Core Package is the center of the software, supporting all the basic functions of the laboratory.
Handle samples and orders on patients and donors from test requests through reporting of results. Look up patient demographics and test history.
Minimum hardware specifications
Required software
Optional software
Minimum hardware specifications
Required software
Minimum hardware specifications
Required software
RVU Software utilization saves time and reduces transcription errors by importing test results directly from the testing analysis software into your HistoTrac Software.
The larger your volume, the more important it is to have the software manage the data entry. With imported results, the review cycle can be reduced, minimizing the need for detailed visual review and freeing time to focus on other tasks. Use an RVU for every testing method.
These interfaces may be bi-directional and begin by exporting a sample ID from HistoTrac to the RVU Software—no more manual entry of sample IDs into text files. After testing is completed and results are analyzed, the user imports test results from the RVU Software into the HistoTrac Software. For a uni-directional interface, the user would only import from the RVU to HistoTrac.
The workflow for the use of RVU is facilitated by batching tests and building trays in your HistoTrac Software. The build-a-tray feature helps you create tray maps organized by various criteria.
HistoTrac Software can be connected with your hospital and/or laboratory system to provide full integration with the patient’s electronic medical record. Patient demographics, test orders, billing transactions, and test results or reports move between medical software systems using an HL7 formatted transaction. Each interface is separate and requires the accompanying hospital or laboratory information system interface.
Validate your HistoTrac database to ensure all patient data is up-to-date and accurate.
This module provides a means to identify differences between the patient data in HistoTrac Software and UNOS. The UNOS Wait List is downloaded from UNet and imported into HistoTrac with validation between the two systems using the patient’s identification.
The user can update HistoTrac values such as the UNOS-listed flag, CPRA, and patient status with data from the Waitlist file, and apply updates to the entire list or individual records.
The UNOS Unacceptables utility provides an API with the patient’s unacceptable specificities, formatted according to the UNET standard. This feature not only saves time but also eliminates transcription errors.
Tests can be ordered for cytotoxicity or flow methods. The donor is added to the patient’s case. A custom-designed report provides test results for coordinators or surgeons.
This module is designed to export donor and/or recipient molecular typing results for Form 117 and Form 22. An API sends the most recent results to the NMDP for import into the registry database on a scheduled basis, perhaps daily, thus saving time and eliminating transcription errors.
The Paired Kidney Exchange (PKE) module provides a complete and flexible tool for swapping previously incompatible patient/donor pairs. The data for calculating matched pairs are all found within the HistoTrac database — no more sending data to third-party programs. Select the optimal chain length, determine the source of antibody testing used to rule out donor incompatibility, and establish the factors that influence the PKE score for each chain.
The PKE module provides the data and programming required for formatting sensitized patients with a compatible donor. As patient test data are updated, the matches can be re-run without sending new data to a third-party system.
The match program provides the flexibility needed to use the criteria of your transplant program. Adjust the parameters for one patient that might be different from another.
The program can be run by the clinical staff or can be used within the laboratory. A report documents the chains created.
Not all transplant programs use the same matching protocol when selecting a donor for a patient. The HistoTrac PKE module provides choices at each of the various match criteria:
Each chain’s score is calculated based on user-defined rankings such as the length of time the patient has been waiting for a transplant, the patient’s antibody level, and the importance of HLA match between the pair members. The higher the score, the more difficult the patient is to transplant.
For Laboratory Use.