Search
Petroleum quality analysis refers to the refining of petroleum crude oil and the processing and purifying of raw natural gas. Products are divided into light distillates (LPG C3-C4), gasoline (C3-C12), naphtha (C6-14), middle distillates (kerosene, diesel C9-C16, jet fuel C9-C17) and heavy distillates (lubricating oil C20+).
Let one of our experts help you tailor a solution specific to your application and workflow needs.
Select from a choice of available products according to the process, matrix, application and analyte of interest.
Upstream | Midstream | Downstream | ||
|---|---|---|---|---|
Natural Gas & Condensates | Energy Content (BTU), DHA, SIMDIS, H2S/Organic Sulfur GC, GC/MS | |||
| Sulfur, Metals ICP-OES | ||||
| F, Cl, S Combustion IC | ||||
Crude Oil | RGA, DHA GC, GC/MS | |||
| Biomarkers GC-HRMS | H2S, CO2, NOX GC | Oxygenates, Aromatics GC, GC/MS | ||
| SIMDIS GC, GC/MS | F, Cl, S Combustion IC | BTEX*, PAHs GC, GC/MS | ||
| S, Metals | PAHs ICP-OES | HPLC, GC | ||||
| LIMS leverages all process steps *benzene, toluene, ethyl benzene, xylenes | ||||