Search
The Thermo Scientific iCAP MX Series ICP-MS comprises both single and triple quadrupole ICP-MS systems and is ideal for applications ranging from high-throughput environmental monitoring to ultra-trace elemental analysis of industrial chemicals. Discover which system is right for your laboratory.
|
|
| Description | |
Offers maximum uptime and productivity for a wide range of sample types with exceptional ease of use. |
Achieves advanced interference removal and supplies the ease of use, performance and flexibility to explore new, innovative applications. |
| Interference removal | |
Qcell collision/reaction cell (CRC) capability—Uses single-mode HeKED interference removal for commonly encountered interferences. Other collision reaction gases can also be used. |
Advanced Qcell collision/reaction cell (CRC) with triple quadrupole capability—Ideal for analyzing samples when isobaric interferences cannot be removed by single quadrupole technology. |
| Typical applications |
|
Analysis of environmental, food safety, pharmaceutical and industrial samples. |
Analysis of samples containing complex interferences, such as those from metallurgy applications or those that support research. |
For Research Use Only. Not for use in diagnostic procedures.