Search
![]() | ![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
| 3 | 488, 532, 561 | 610/20 | 565 | 606 | (in buffer) 1 | flow cytometry |
Invitrogen PE-eFluor 610 dye is a high-performance tandem fluorophore designed for use with flow cytometers equipped with blue (488 nm), green (532 nm), or yellow-green (561 nm) laser. It has a peak emission of 607 nm that can be detected using typical filters for PE-Texas Red dye, such as a 610/20 bandpass filter.
We offer PE-eFluor 610 dye conjugated to a variety of antibodies, and amplification substrates optimized for cellular labeling and detection.
Search PE-eFluor 610 primary antibodies Search PE-eFluor 610 secondary antibodies
Comparison of staining performance of PE-eFluor 610 to PE-Texas Red dye. Normal human peripheral blood cells (PBMCs) were stained with CD3 Monoclonal Antibody (UCHT1), PE-eFluor 610 (dark orange line) or CD3 Monoclonal Antibody (7D6), PE-Texas Red (light orange line).
Protocols that fit your needs in flow cytometry ranging from sample preparation to numerous cell stimulation conditions, staining, immunophenotyping, and data analysis strategies.
A handy reference poster featuring the broad range of our available dyes and labeling reagents.
Protocols that fit your needs in imaging ranging from sample and assay preparation to staining, labeling, and data analysis strategies.
A tool for selecting the optimal fluorescent dyes for your Invitrogen EVOS cell imaging systems.
Cy™ is a trademark or registered trademark of GE Healthcare.
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and are used under license.
For Research Use Only. Not for use in diagnostic procedures.