Search
CloneMiner II cDNA Library Construction Kits enable rapid construction of highly representative cDNA libraries. Gateway recombination cloning avoids restriction digestion and inefficient ligation reactions, making library construction faster and offering more complete cDNA representation. The high efficiency SuperScript III reverse transcriptase is included, enhancing the possibility that users may discover previously unobtainable, fully intact clones.
CloneMiner II Kits include SuperScript III RT, Gateway vectors, reagents for cDNA library construction and ElectroMax DH10B T1-Phage-resistant Competent Cells.
Sufficient reagents are supplied for 5 cDNA library construction reactions.
For Research Use Only. Not for use in diagnostic procedures.