Search
Learn about PCR and qPCR plastics and plate seals, what to look for when deciding which to use, and the impact they can have on the quality of DNA amplification studies. Browse the FAQs on PCR plates and PCR tubes; find resources to support your PCR and qPCR efforts with a focus on PCR plastics.
Ask yourself these questions to help determine which type(s) of PCR plastics you should use.
View tips on tackling PCR/qPCR issues potentially related to PCR consumables.
PCR (Polymerase Chain Reaction) plates are specialized microplates designed for use in molecular biology research. They typically contain an array of 96 or 384 wells made of polypropylene, where each well holds an individual PCR or other reaction involving nucleic acids.
PCR plates are primarily used for standard PCR reactions, which involve amplifying DNA through a series of heating and cooling cycles. On the other hand, qPCR plates are used for quantitative PCR, which measures the amount of DNA present in a sample in real time. qPCR plate wells are typically made of white or frosted polypropylene to reduce background fluorescence, which can interfere with accurate detection of the target nucleic acid.
Processing of multiple samples in parallel is easier in a PCR plate compared to individual tubes, saving sample preparation time and helping reduce user fatigue caused by capping and de-capping PCR tubes. Additionally, plates are more compatible with automated liquid handling systems, enabling further increases in throughput of PCR and qPCR workflows.
PCR and qPCR plates are well suited for many DNA and RNA applications including DNA sequencing, genotyping, and gene expression analysis. The versatility of these plates helps make them an essential item for molecular biology laboratories.
PCR and qPCR plates are recommended for single use only. Re-use of plates heightens the risk of cross-contamination and unreliable results. If you routinely use less than half a plate, consider options such as segmented PCR plates and PCR strip tubes.
Explore the features of Thermo Scientific PCR and qPCR plates
Search the FAQ database for more specific information about PCR plates
PCR tubes are small, coned-shaped vessels made of polypropylene and used in Polymerase Chain Reaction (PCR) experiments. PCR tubes are designed to avoid evaporation of the PCR reaction mixture (comprising DNA templates, primers, nucleotides, and polymerase enzymes) and withstand repeated temperature fluctuations that occur during the PCR reaction.
PCR strips, aka PCR tube strips, typically consist of 8 or 12 PCR tubes joined together in a linear arrangement. A PCR strip may have attached caps or utilize a strip of caps for tube sealing.
PCR strips provide a convenient and efficient format for conducting multiple PCR reactions in parallel, making them a popular choice in molecular biology laboratories. The use of PCR tube strips facilitates handling and organization of the samples as well as simultaneous processing of multiple samples, helping save time and reducing labor. PCR strips compatible with standard thermal cyclers can be easily placed into the instrument's block for thermal cycling.
Search the FAQ database for more specific information about PCR tubes
Explore the features of Thermo Scientific PCR tubes and strip tubes
Find the right plastic consumables that are compatible with your thermal cyclers and qPCR instruments.
Read articles on how to maximize workflow success with excellent plastics.