Search
TaqMan™ Assays are probe and primer sets based on 5’-nuclease assay chemistry using TaqMan™ MGB (minor groove binder) probes, the most quantitative expression technology available.
Choose from a variety of primer and probe sets (assays) designed by Thermo Fisher scientists based on format and research area.
| What assay format do you want? | What type of research are you doing? | Use this TaqMan Assay/Array |
|---|---|---|
| Single tube | Mature miRNA detection | TaqMan Advanced miRNA Assays |
| Pri-miRNA detection | TaqMan Pri-miRNA Assays | |
| Long non-coding RNA detection | TaqMan Non-coding RNA Assays | |
| 96-well plate | Mature miRNA detection | TaqMan Advanced miRNA Plates |
| 384-well microfluidic card | Mature miRNA detection, fixed content | TaqMan miRNA Array Cards |
| Mature miRNA detection, custom content | Custom TaqMan miRNA Array Cards | |
| OpenArray plate | Large-scale profiling (human/rodent) | TaqMan OpenArray MicroRNA Panels |
Submit your own sequences and we will make TaqMan Assays to your specifications.
| What assay format do you want? | Use this TaqMan Assay/Array |
|---|---|
| Single tube | Custom TaqMan Small RNA Assays |
TaqMan Assays are available in the following formats:
For Research Use Only. Not for use in diagnostic procedures.