Germanium • Metalloid
Primary XPS region: Ge2p, Ge3d
Overlapping regions: W4f, F2s
Binding energies of common chemical states:
Chemical stateBinding energy Ge2p3/2Binding energy Ge3d5/2
Ge elemental1217.3 eV29.3 eV
GeO1218.0 eV30.9 eV
GeO21220.2 eV32.5 eV
Experimental information
  • Ge2p or Ge3d are typically used.
    • Ge3d may be preferable if germanium is buried and at low concentration.
      • Ge3d electrons have higher kinetic energy and therefore have a deeper sampling depth compared to the low kinetic energy Ge2p electrons.
Interpretation of XPS spectra
  • Ge2p region has significantly split spin-orbit components (Δ=31.1eV, intensity ratio=0.478).
    • Ge2p peaks have symmetric lineshape for elemental germanium and its compounds.
    • Typically only the Ge2p3/2 component of the Ge2p region is fitted and quantified.
    • Loss feature may be observed between spin-orbit components (1,235eV) and to higher binding energy of Ge2p1/2 component (1,265eV).
Ge2p Spectrum Native Oxide
Germanium • Non-Metals
  • Ge3d peak has overlapping spin-orbit components (Δ=0.58eV, intensity ratio=0.67).
    • Splitting is observable for elemental germanium, but may be ignored for germanium compounds.
    • Ge3d peaks have symmetric lineshape for elemental germanium and its compounds.
 Ge3d Spectrum Native Oxide
General comments
  • Ge LMM peaks (between 300eV-600eV) overlap with many XPS peaks from other elements.
Element Crystal FCC
About this element
Symbol: Ge
Date of discovery: 1886
Name origin: Latin Germania
Appearance: grayish
Discoverer: Clemens Winkler
Obtained from: copper, zinc, lead
Neutrons: 41
Shell structure: 2,8,18,4
Electron configuration: [Ar]3d104s24p2
Melting point: 1211.4 K
Boiling point: 3093 K
Density[kg/m3]: 5323
Molar volume: 13.63 × 10-6 m3/mol
Oxidation state: 4
Protons/electrons: 32
Crystal structure: cubic face centered
Chemically similar to tin, germanium is an important semiconductor. Due to its small band gap, germanium is highly sensitive to infrared light and is used as detector material in infrared and Raman spectrometers. Germanium’s refraction and dispersion properties make it useful in wide-angle camera lenses and in microscope objective lenses. Silicon germanide (SiGe) is becoming an important material in high-speed integrated circuits. Germanium transistors are used in modern electric guitars to simulate the sound of Rock and Roll-era amplifiers.
Products
Nexsa G2 XPS
  • 미세 초점 X-선 소스
  • 독자적인 다중기법 옵션
  • 단일 원자 & 클러스터 이온 깊이 프로파일링을 위한 듀얼 모드 이온 소스
기기 카드 원본 스타일시트
K-Alpha XPS
  • 고분해능 XPS
  • 빠르고 효율적이며 자동화된 워크플로우
  • 깊이 프로파일링을 위한 이온 소스
ESCALAB Xi+ XPS
  • 높은 스펙트럼 분해능
  • 다중 기법 표면 분석
  • 광범위한 시료 준비 및 확장 옵션
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
재품 표 사항에 대한 스타일시트
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
재품 표 사항에 대한 스타일시트
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
재료 과학을 위한 전자 현미경
서비스
최적의 시스템 성능을 보장하기 위해 세계 수준의 현장 서비스 전문가 네트워크, 기술 지원 및 인증된 예비 부품을 제공해드립니다.