Search
Western blot analysis from lysates of MCF-7 human breast cancer cell line. PVDF membrane was probed with 1 µg/mL of human AFAP Antigen Affinity-purified Polyclonal Antibody (Product # PA5-47636) followed by HRP-conjugated Anti-Goat IgG Secondary Antibody. A specific band was detected for AFAP at approximately 110 kDa (as indicated).